EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Antioxidants Inhibit Formation of 3-Monochloropropane-1,2-diol Esters in Model Reactions.
PMID 26478126
Abstract
The capacities of six antioxidants to inhibit the formation of 3-monochloropropane-1,2 diol (3-MCPD) esters were examined in this study. Inhibitory capacities of the antioxidants were investigated both in chemical models containing the precursors (tripalmitoyl glycerol, 1,2-dipalmitoyl-sn-glycerol, monopalmitoyl glycerol, and sodium chloride) of 3-MCPD esters and in oil models (rapeseed oil and sodium chloride). Six antioxidants, butylated hydroxytoluene (BHT), butylated hydroxy anisole (BHA), tert-butyl hydroquinone (TBHQ), propyl gallate (PG), L-ascorbyl palmitate (AP), and α-tocopherol (VE), were found to exhibit inhibiting capacities on 3-MCPD ester formation both in chemical models and in oil models. TBHQ provided the highest inhibitory capacity both in chemical models and in oil models; 44% of 3-MCPD ester formation was inhibited in the presence of TBHQ (66 mg/kg of oil) after heating of rapeseed oil at 230 °C for 30 min, followed by PG and AP. BHT, BHA, and VE appeared to have weaker inhibitory abilities in both models. VE exhibited the lowest inhibition rate; 22% of 3-MCPD esters were inhibited in the presence of VE (172 mg/kg of oil) after heating of rapeseed oil at 230 °C for 30 min. In addition, the inhibition rates of PG and VE decreased dramatically with an increase in temperature or heating time. The results suggested that some antioxidants, such as TBHQ, PG, and AP, could be the potential inhibitors of 3-MCPD esters in practice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9135
1,2-Dipalmitoyl-sn-glycerol, ≥99%
C35H68O5
M65807
2-Methylpentane, ≥99%
C6H14
68312
2-Methylpentane, ≥95.0% (GC)
C6H14
442360
2-Methylpentane, analytical standard
C6H14
M1640
DL-α-Palmitin, ≥99%
C19H38O4
T5888
Glyceryl tripalmitate, ≥99%
C51H98O6
T8127
Glyceryl tripalmitate, ≥85%
C51H98O6
296317
Hexadecane, anhydrous, ≥99%
C16H34
442679
Hexadecane, analytical standard
C16H34
H6703
Hexadecane, ReagentPlus®, 99%
C16H34
33352
Methanolic Base, 0.5 M CH3ONa (0.5N), for GC derivatization
CH3NaO
33080
Methanolic Base, 0.5 M CH3ONa (0.5N), for GC derivatization
CH3NaO
P20009
Phenylboronic acid, 95%
C6H7BO2
746401
Sodium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
BrNa
793574
Sodium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
BrNa
S4547
Sodium bromide, BioXtra, ≥99.0%
BrNa
71329
Sodium bromide, BioUltra, ≥99.5% (AT)
BrNa
02119
Sodium bromide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance)
BrNa
460591
Sodium bromide, anhydrous, beads, −10 mesh, 99.999% trace metals basis
BrNa
229881
Sodium bromide, ≥99.99% trace metals basis
BrNa
310506
Sodium bromide, ACS reagent, ≥99.0%
BrNa
220345
Sodium bromide, ReagentPlus®, ≥99%
BrNa
164992
Sodium methoxide, reagent grade, 95%, powder
CH3NaO
403067
Sodium methoxide solution, ACS reagent, 0.5 M CH3ONa in methanol (0.5N)
CH3NaO
156256
Sodium methoxide solution, 25 wt. % in methanol
CH3NaO