EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Influence of pyrolysis temperature on characteristics and phosphate adsorption capability of biochar derived from waste-marine macroalgae (Undaria pinnatifida roots).
PMID 26482944
Abstract
The collected roots of Undaria pinnatifida, the main waste in farming sites, accounting for 40-60% of annual production, was pyrolyzed under temperature ranging from 200 to 800°C to evaluate the influence of pyrolysis temperature on biochar properties and phosphate adsorption capacity. It was confirmed that an increase in the pyrolysis temperature led to a decrease of the yield of biochar, while ash content remained almost due to carbonization followed by mineralization. Elemental analysis results indicated an increase in aromaticity and decreased polarity at a high pyrolysis temperature. When the pyrolysis temperature was increased up to 400°C, the phosphate adsorption capacity was enhanced, while a further increase in the pyrolysis temperature lowered the adsorption capacity due to blocked pores in the biochar during pyrolysis. Finally, a pot experiment revealed that biochar derived from waste-marine macroalgae is a potent and eco-friendly alternative material for fertilizer after phosphate adsorption.