EMAIL THIS PAGE TO A FRIEND
Journal of biomedical nanotechnology
Novel IL-2-Poly(HPMA)Nanoconjugate Based Immunotherapy.
PMID 26485935
Abstract
Interleukin-2 (IL-2) possesses a strong stimulatory activity for activated T and NK cells and it is an attractive molecule for immunotherapy. Nevertheless, extremely short half-life and severe toxicities associated with high-dose IL-2 treatment are serious and limiting drawbacks. In order to increase IL-2 half-life in vivo, we covalently conjugated synthetic semitelechelic polymeric carrier based on N-(2-hydroxypropyl)methacrylamide (HPMA) to IL-2. Thus, we synthesized IL-2-poly(HPMA) conjugate containing 2-3 polymer chains per IL-2 molecule in average. Such conjugate has lower biologic activity in comparison to IL-2 in vitro. However, it exerts much higher activity than IL-2 in vivo as shown by expansion of memory CD8+ T, NK, NKT, γδT and Treg cells. Moreover, IL-2-poly(HPMA) extremely effectively potentiates CD8+ T cell peptide-based vaccination. IL-2-poly(HPMA) shows also much longer half-time in circulation than IL-2 (-4 h versus -5 min). Collectively, modification of IL-2 with poly(HPMA) chains dramatically improves its potency and pharmacologic features in vivo, which have implications for immunotherapy. To our knowledge, this is the first proof-of-concept report of the use of polymer/protein modification of IL-2 to obtain more pronounced biological activity.