EMAIL THIS PAGE TO A FRIEND
Neural regeneration research
Differentiation of Wharton's jelly mesenchymal stem cells into neurons in alginate scaffold.
PMID 26487861
Abstract
Alginate scaffold has been considered as an appropriate biomaterial for promoting the differentiation of embryonic stem cells toward neuronal cell lineage. We hypothesized that alginate scaffold is suitable for culturing Wharton's jelly mesenchymal stem cells (WJMSCs) and can promote the differentiation of WJMSCs into neuron-like cells. In this study, we cultured WJMSCs in a three-dimensional scaffold fabricated by 0.25% alginate and 50 mM CaCl2 in the presence of neurogenic medium containing 10 μM retinoic acid and 20 ng/mL basic fibroblast growth factor. These cells were also cultured in conventional two-dimensional culture condition in the presence of neurogenic medium as controls. After 10 days, immunofluorescence staining was performed for detecting β-tubulin (marker for WJMSCs-differentiated neuron) and CD271 (motor neuron marker). β-Tubulin and CD271 expression levels were significantly greater in the WJMSCs cultured in the three-dimensional alginate scaffold than in the conventional two-dimensional culture condition. These findings suggest that three-dimensional alginate scaffold cell culture system can induce neuronal differentiation of WJMSCs effectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1306228
ANTI-CD271(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
GW22755 Anti-CD44 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405590
Anti-CD44 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA005785
Anti-CD44 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300691
Anti-CD44 antibody produced in rabbit, affinity isolated antibody
GW22756 Anti-Endoglin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
E7534
Anti-Endoglin antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
AV33068
Anti-ENG (AB1) antibody produced in rabbit, IgG fraction of antiserum
HPA011862
Anti-ENG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
N3908
Anti-Nerve Growth Factor Receptor (NGFR p75) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2500694
Anti-NGFR antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502011
Anti-NGFR antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA004765
Anti-NGFR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500298
Anti-NGFR antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2501275
Anti-THY1 antibody produced in goat, affinity isolated antibody
HPA003733
Anti-THY1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
V1514
Anti-VE-Cadherin (CD144) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
46955
Fluorescein, for fluorescence, free acid
C20H12O5
F0291 hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
SAB4700258 Monoclonal Anti-CD105 antibody produced in mouse, clone MEM-226, purified immunoglobulin, buffered aqueous solution
SAB4700262 Monoclonal Anti-CD105 antibody produced in mouse, clone MEM-229, purified immunoglobulin, buffered aqueous solution
C7923 Monoclonal Anti-CD44 antibody produced in mouse, clone A3D8, purified immunoglobulin, buffered aqueous solution
SAB1402714
Monoclonal Anti-CD44 antibody produced in mouse, clone 1E1, purified immunoglobulin, buffered aqueous solution
SAB4700179 Monoclonal Anti-CD44 antibody produced in mouse, clone MEM-85, purified immunoglobulin, buffered aqueous solution
SAB4700184 Monoclonal Anti-CD44 antibody produced in mouse, clone MEM-263, purified immunoglobulin, buffered aqueous solution
SAB4700188 Monoclonal Anti-CD44 antibody produced in rat, clone IM7, purified immunoglobulin, buffered aqueous solution
WH0002022M1
Monoclonal Anti-ENG antibody produced in mouse, clone 4C11, purified immunoglobulin, buffered aqueous solution
AMAB90925
Monoclonal Anti-ENG antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1912, purified immunoglobulin, buffered aqueous glycerol solution
N5408 Monoclonal Anti-Nerve Growth Factor Receptor (NGFR p75) antibody produced in mouse, clone ME20.4, ascites fluid
SAB5300237
Monoclonal Anti-NGFR antibody produced in mouse, clone 2F1C2, ascites fluid