EMAIL THIS PAGE TO A FRIEND
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
sPLA2-IIA Augments Oxidized LDL-Induced MCP-1 Expression in Vitro Through Activation of Akt.
PMID 26488172
Abstract
Group IIA secretory phospholipase A2 (sPLA2-IIA) has an important role in atherosclerosis. In this study, we explored whether sPLA2-IIA overexpression could promote atherosclerosis in normal environment alone or with other inflammatory factors. Human aortic smooth muscle cells (HASMCs) were transduced with Lv-GFP-sPLA2-IIA, a plasmid containing sPLA2-IIA coupled with green fluorescent protein (GFP). Cells were incubated in the presence or absence of oxidized low-density lipoprotein (LDL), sPLA2 inhibitor LY315920 or PI3K/Akt inhibitor LY294002. The mRNA expression and protein secretion of monocyte chemoattractant protein-1 (MCP-1) were assessed by quantitative real-time polymerase chain reaction (QRT-PCR) and enzyme-linked immunosorbent assay (ELISA), respectively. Phosphorylation of Akt was examined by western blotting. Lv-GFP-sPLA2-IIA-transduced HASMCs remained fluorescent during 72 h of the study period with infection ratio of around 80%. The mRNA expression and protein secretion of MCP-1 was not altered in groups of HASMCs, Lv-GFP transduced and Lv-GFP-sPLA2-IIA-transduced HASMCs (p>0.05), but was significantly increased in the presence of oxidized LDL especially in Lv-GFP-sPLA2-IIA transduction group (p<0.01). However, with the addition of LY315920, this enhancement was notably decreased (p<0.05). This enhancement was also markedly abolished by co-incubation with LY294002, paralleled with suppressed Akt phosphorylation. Overexpression of sPLA2-IIA does not alter MCP-1 level at baseline, but could enhance the atherogenic effect of oxidized LDL in HASMCs, at least partly due to activation of Akt. These findings may provide a strategy for treatment of inflammatory cardiovascular diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3700864 Anti-Rabbit IgG (Fc specific)-Alkaline Phosphatase antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3700865 Anti-Rabbit IgG (Fc specific)-Biotin antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700862 Anti-Rabbit IgG (Fc specific)-Fluorescein antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700863 Anti-Rabbit IgG (Fc specific)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700860 Anti-Rabbit IgG (Fc specific)-Rhodamine antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700866 Anti-Rabbit IgG (Fc specific)-Texas Red® antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700979 Anti-Rabbit IgG (H+L), F(ab) fragment-Biotin antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700977 Anti-Rabbit IgG (H+L), F(ab) fragment-Fluorescein antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700978 Anti-Rabbit IgG (H+L), F(ab) fragment-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700975 Anti-Rabbit IgG (H+L), F(ab) fragment-Rhodamine antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700980 Anti-Rabbit IgG (H+L), F(ab) fragment-Texas Red® antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB4600003 Anti-Rabbit IgG (H+L), highly cross-adsorbed, Biotin antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600010
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 350 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600036
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 488A antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600061
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 555 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600076
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 568 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600099
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 594 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600132
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 633 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600155
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 640R antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600177
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 647 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600208
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 680R antibody produced in donkey, ~2 mg/mL, affinity isolated antibody
SAB4600372 Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF750 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600373 Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF750 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB3700935 Anti-Rabbit IgG (H+L)-Alkaline Phosphatase antibody produced in donkey, affinity isolated antibody, buffered aqueous solution
SAB3700936 Anti-Rabbit IgG (H+L)-Biotin antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700933 Anti-Rabbit IgG (H+L)-Fluorescein antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700934 Anti-Rabbit IgG (H+L)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700925 Anti-Rabbit IgG (H+L)-Rhodamine antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700937
Anti-Rabbit IgG (H+L)-Texas Red® antibody produced in donkey, affinity isolated antibody, lyophilized powder