EMAIL THIS PAGE TO A FRIEND
General physiology and biophysics
The cytoprotective effects of ethanol extract of Ecklonia cava against oxidative stress are associated with upregulation of Nrf2-mediated HO-1 and NQO-1 expression through activation of the MAPK pathway.
PMID 26492067
Abstract
The aim of the present study was to examine the cytoprotective effect of Ecklonia cava against oxidative stress in C2C12 myoblasts. The ethanol extract of E. cava (EEEC) prevented hydrogen peroxide (H₂O₂)-induced inhibition of the growth of C2C12 myoblasts and exhibited scavenging activity against intracellular reactive oxygen species (ROS) induced by H₂O₂. EEEC treatment attenuated H2O2-induced comet tail formation and phospho-histone γH2A.X expression. Furthermore, EEEC treatment enhanced the level of the phosphorylated form of nuclear factor erythroid 2- related factor 2 (Nrf2) and its nuclear translocation, which was associated with the induction of heme oxygenase-1 (HO-1) and NADPH-quinone oxidoreductase 1 (NQO-1). Zinc protoporphyrin IX, a HO-1 competitive inhibitor, significantly abolished the protective effects of EEEC against H₂O₂-induced ROS generation and growth inhibition in C2C12 myoblasts. Transient transfection with Nrf2-specific small interfering RNA restored the elevated HO-1 and NQO-1 expression and the phosphorylation of Nrf2 to near normal levels. The EEEC treatment also induced the activation of mitogen-activated protein kinases (MAPKs), and specific inhibitors of MAPKs abolished upregulated HO-1 and NQO-1, as well as the phosphorylation of Nrf2. Taken together, these data suggest that EEEC attenuates oxidative stress by activating Nrf2-mediated HO-1 and inducing NQO-1 via the activation of MAPK signaling pathways.