EMAIL THIS PAGE TO A FRIEND
International journal of molecular sciences
MyoD Is a Novel Activator of Porcine FIT1 Gene by Interacting with the Canonical E-Box Element during Myogenesis.
PMID 26492245
Abstract
Fat-induced transcript 1 (FIT1/FITM1) gene is a member of the conserved gene family important for triglyceride-rich lipid droplet accumulation. FIT1 gene displays a similar muscle-specific expression across pigs, mice, and humans. Thus pigs can act as a useful model of many human diseases resulting from misexpression of FIT1 gene. Triglyceride content in skeletal muscle plays a key role in pork meat quality and flavors. An insertion/deletion mutation in porcine FIT1 coding region shows a high correlation with a series of fat traits. To gain better knowledge of the potential role of FIT1 gene in human diseases and the correlations with pork meat quality, our attention is given to the region upstream of the porcine FIT1 coding sequence. We cloned ~1 kb of the 5'-flanking region of porcine FIT1 gene to define the role of this sequence in modulating the myogenic expression. A canonical E-box element that activated porcine FIT1 promoter activity during myogenesis was identified. Further analysis demonstrated that promoter activity was induced by overexpression of MyoD1, which bound to this canonical E-box during C2C12 differentiation. This is the first evidence that FIT1 as the direct novel target of MyoD is involved in muscle development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA021289
Anti-BBS9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408674
Anti-BBS9 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1304992 ANTI-DIO1 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1407254
Anti-LMOD1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101354
Anti-LMOD1 antibody produced in rabbit, affinity isolated antibody
SAB4200200
Anti-LMOD1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
HPA028325
Anti-LMOD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA028435
Anti-LMOD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
HPA030097
Anti-LMOD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab3
SAB2107079
Anti-LMOD1 antibody produced in rabbit, affinity isolated antibody
SAB2501587
Anti-MYOG (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA038093
Anti-MYOG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410814
Anti-MYOG antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
M9818 Anti-Myopodin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305785
ANTI-PTHB1(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA030665
Anti-SYNPO2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500586
Anti-SYNPO2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
EHU049551 MISSION® esiRNA, esiRNA human SYNPO2 (esiRNA1)
EHU138681 MISSION® esiRNA, esiRNA human MYOG (esiRNA1)
EMU152131 MISSION® esiRNA, esiRNA targeting mouse Synpo2
SAB1402173 Monoclonal Anti-DIO1 antibody produced in mouse, clone 1E4, purified immunoglobulin, buffered aqueous solution
SAB4200496
Monoclonal Anti-LMOD1 antibody produced in mouse, ~1.0 mg/mL, clone LMOD1-12, purified immunoglobulin
SAB1401605
Monoclonal Anti-LMOD1 antibody produced in mouse, clone 3B8, purified immunoglobulin, buffered aqueous solution
WH0004656M1
Monoclonal Anti-MYOG antibody produced in mouse, clone 2B7, purified immunoglobulin, buffered aqueous solution
SAB1404106
Monoclonal Anti-MYOG antibody produced in mouse, clone 2E7, purified immunoglobulin, buffered aqueous solution
SAB1409339
Monoclonal Anti-MYOG antibody produced in mouse, clone 3E3, purified immunoglobulin, buffered aqueous solution
M5815 Monoclonal Anti-Myogenin antibody produced in mouse, clone F12B, purified immunoglobulin, buffered aqueous solution