EMAIL THIS PAGE TO A FRIEND
Pancreas
Interplay Between Gemcitabine and Erlotinib Over Pancreatic Adenocarcinoma Cells.
PMID 26495790
Abstract
Pancreatic ductal adenocarcinoma remains as a chemoresistant disease with the poorest prognosis. Gemcitabine has been the standard treatment during the last decade. Erlotinib, a tyrosine kinase inhibitor, in combination with gemcitabine produces a small increase in survival. However, these results remain insufficient. The aim of this study was to investigate the molecular interplay in vitro between them regarding their effects over cytotoxicity, proliferation, apoptosis, and invasion. Using the human pancreatic cancer cell lines Panc-1 and BxPC-3 in vitro, the effects of gemcitabine and erlotinib therapy on growth, proliferation, and invasion were tested by cytotoxicity, cell cycle, and Annexin V-Fluorescein Isothiocyanate analysis, reverse transcription polymerase chain reaction, protein expression, and Chip assays. Therapy decreased cell proliferation causing G0/G1 phase cell cycle arrest with induction of apoptosis in the Panc-1 cell line. This blockade was associated with increased p27 expression. Besides, treatments enhanced the nuclear factor-κB (NF-κB) pathway and the binding of NF-κB to the promoters of genes related to the proliferation and the evasion of apoptosis. Our data suggest that, although gemcitabine and erlotinib exert antiproliferative effects over pancreatic cancer cell lines, the gemcitabine-induced activation of NF-κB expression and its DNA-binding activities are important drawbacks of this treatment against pancreatic cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS022564
Erlotinib hydrochloride
C22H24ClN3O4