EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Determination of Apparent Amylose Content in Rice by Using Paper-Based Microfluidic Chips.
PMID 26495809
Abstract
Determination of apparent amylose content in rice is a key function for rice research and the rice industry. In this paper, a novel approach with paper-based microfluidic chip is reported to determine apparent amylose content in rice. The conventional color reaction between amylose and iodine was employed. Blue color of amylose-iodine complex generated on-chip was converted to gray and measured with Photoshop after the colored chip was scanned. The method for preparation of the paper chip is described. In situ generation of iodine for on-chip color reaction was designed, and factors influencing color reaction were investigated in detail. Elimination of yellow color interference of excess iodine by exploiting color removal function of Photoshop was presented. Under the optimized conditions, apparent amylose content in rice ranging from 1.5 to 26.4% can be determined, and precision was 6.3%. The analytical results obtained with the developed approach were in good agreement with those with the continuous flow analyzer method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
104817
Trichloro(octadecyl)silane, ≥90%
C18H37Cl3Si