EMAIL THIS PAGE TO A FRIEND
PLoS genetics
Protein O-Glucosyltransferase 1 (POGLUT1) Promotes Mouse Gastrulation through Modification of the Apical Polarity Protein CRUMBS2.
PMID 26496195
Abstract
Crumbs family proteins are apical transmembrane proteins with ancient roles in cell polarity. Mouse Crumbs2 mutants arrest at midgestation with abnormal neural plate morphology and a deficit of mesoderm caused by defects in gastrulation. We identified an ENU-induced mutation, wsnp, that phenocopies the Crumbs2 null phenotype. We show that wsnp is a null allele of Protein O-glucosyltransferase 1 (Poglut1), which encodes an enzyme previously shown to add O-glucose to EGF repeats in the extracellular domain of Drosophila and mammalian Notch, but the role of POGLUT1 in mammalian gastrulation has not been investigated. As predicted, we find that POGLUT1 is essential for Notch signaling in the early mouse embryo. However, the loss of mouse POGLUT1 causes an earlier and more dramatic phenotype than does the loss of activity of the Notch pathway, indicating that POGLUT1 has additional biologically relevant substrates. Using mass spectrometry, we show that POGLUT1 modifies EGF repeats in the extracellular domain of full-length mouse CRUMBS2. CRUMBS2 that lacks the O-glucose modification fails to be enriched on the apical plasma membrane and instead accumulates in the endoplasmic reticulum. The data demonstrate that CRUMBS2 is the target of POGLUT1 for the gastrulation epithelial-to-mesenchymal transitions (EMT) and that all activity of CRUMBS2 depends on modification by POGLUT1. Mutations in human POGLUT1 cause Dowling-Degos Disease, POGLUT1 is overexpressed in a variety of tumor cells, and mutations in the EGF repeats of human CRUMBS proteins are associated with human congenital nephrosis, retinitis pigmentosa and retinal degeneration, suggesting that O-glucosylation of CRUMBS proteins has broad roles in human health.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4502019
Anti-NOTCH1 (Cleaved-Val1754) antibody produced in rabbit, affinity isolated antibody
N4788 Anti-NOTCH1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2101618
Anti-NOTCH1 antibody produced in rabbit, affinity isolated antibody
SAB2701800
Anti-sox2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
S9072 Anti-SOX2 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
AV38232
Anti-SOX2 antibody produced in rabbit, IgG fraction of antiserum
SAB3500363
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104660
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody
SAB4300669
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody
SAB2701973
Anti-SOX2 antibody produced in rabbit
SAB2701974
Anti-SOX2 antibody produced in rabbit
SAB2103958
Anti-SOX2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4700742 Monoclonal Anti-Notch 1 antibody produced in mouse, clone mN1A, purified immunoglobulin, buffered aqueous solution
SAB4200024 Monoclonal Anti-Notch 1 antibody produced in rat, ~1.0 mg/mL, clone ICN 5B5, purified immunoglobulin, buffered aqueous solution
N6786
Monoclonal Anti-Notch1 antibody produced in mouse, clone mN1A, purified immunoglobulin, buffered aqueous solution
SAB3300078
Monoclonal Anti-Notch1 antibody produced in mouse, clone 3E12, purified immunoglobulin, buffered aqueous glycerol solution
SAB1402292
Monoclonal Anti-NOTCH1 antibody produced in mouse, clone 4G1, purified immunoglobulin, buffered aqueous solution
S1451
Monoclonal Anti-Sox2 antibody produced in mouse, ~1.0 mg/mL, clone SOX2-6, purified immunoglobulin, buffered aqueous solution
SAB5300177
Monoclonal Anti-SOX2 antibody produced in mouse, clone 10F10, ascites fluid
SAB5300369 Monoclonal Anti-SOX2 antibody produced in mouse, clone 10F10C9, ascites fluid
PLA0222
Rabbit anti-Notch1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.