EMAIL THIS PAGE TO A FRIEND
Comptes rendus biologies
ISSR-based analysis of genetic diversity among sorghum landraces growing in some parts of Saudi Arabia and Yemen.
PMID 26496867
Abstract
Inter simple sequence repeats (ISSR) analysis was used to determine the genetic diversity among 15 genotypes of sorghum [Sorghum bicolor (L.) Moench] growing in some parts of Saudi Arabia and Yemen. A total of 92 alleles were amplified, with an average of 13 ISSR alleles per primer. Cluster analysis divided the 15 genotypes into two main groups. Group A consisted of five genotypes with white grains from Jazan and Abha with a similarity coefficient range of 0.527 to 0.818. Group B was comprised of 10 genotypes; two genotypes from Al-Qassim were clearly delimited from the remaining eight samples with a coefficient range from 0.709 to 0.490. The eight genotypes were divided into two clusters; one was comprised of landraces with dark grains from Abha in Saudi Arabia and Ab in Yemen, with a similarity coefficient range between 0.563 and 0.781, and the other cluster was differentiated into three white-colored-grain genotypes and one colored-grain genotype; all samples from North Yemen had a similarity coefficient range from 0.454 to 0.800. The current results encourage further collection and authentication of sorghum landraces in the gene banks of Saudi Arabia.