EMAIL THIS PAGE TO A FRIEND
Oncotarget
miR-149 represses metastasis of hepatocellular carcinoma by targeting actin-regulatory proteins PPM1F.
PMID 26498692
Abstract
microRNAs have been implicated in hepatocellular carcinoma (HCC) metastasis, which is predominant cause of high mortality in these patients. Although an increasing body of evidence indicates that miR-149 plays an important role in the growth and metastasis of multiple types of cancers, its role in the progression of HCC remains unknown. Here, we demonstrated that miR-149 was significantly down-regulated in HCC, which was correlated with distant metastasis and TNM stage with statistical significance. A survival analysis showed that decreased miR-149 expression was correlated with a poor prognosis of HCC as well. We found that over-expression of miR-149 suppressed migration and invasion of HCC cells in vitro. In addition, we identified PPM1F (protein phosphatase, Mg(2+)/Mn(2+)-dependent, 1F) as a direct target of miR-149 whose expression was negatively correlated with the expression of miR-149 in HCC tissues. The re-expression of PPM1F rescued the miR-149-mediated inhibition of cell migration and invasion. miR-149 regulated formation of stress fibers to inhibit migration, and re-expression of PPM1F reverted the miR-149-mediated loss of stress fibers. Moreover, we demonstrated that over-expression of miR-149 reduced pMLC2, a downstream effector of PPM1F, in MHCC-97H cells. In vivo studies confirm inhibition of HCC metastasis by miR-149. Taken together, our findings indicates that miR-149 is a potential prognostic biomarker of HCC and that the miR-149/PPM1F regulatory axis represents a novel therapeutic target for HCC treatment.
Related Materials