EMAIL THIS PAGE TO A FRIEND
Nature biotechnology
In vivo characterization of the physicochemical properties of polymer-linked TLR agonists that enhance vaccine immunogenicity.
PMID 26501954
Abstract
The efficacy of vaccine adjuvants such as Toll-like receptor agonists (TLRa) can be improved through formulation and delivery approaches. Here, we attached small molecule TLR-7/8a to polymer scaffolds (polymer-TLR-7/8a) and evaluated how different physicochemical properties of the TLR-7/8a and polymer carrier influenced the location, magnitude and duration of innate immune activation in vivo. Particle formation by polymer-TLR-7/8a was the most important factor for restricting adjuvant distribution and prolonging activity in draining lymph nodes. The improved pharmacokinetic profile by particulate polymer-TLR-7/8a was also associated with reduced morbidity and enhanced vaccine immunogenicity for inducing antibodies and T cell immunity. We extended these findings to the development of a modular approach in which protein antigens are site-specifically linked to temperature-responsive polymer-TLR-7/8a adjuvants that self-assemble into immunogenic particles at physiologic temperatures in vivo. Our findings provide a chemical and structural basis for optimizing adjuvant design to elicit broad-based antibody and T cell responses with protein antigens.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PP2388 Bacterial 10His Tag Vector Set, plasmid vectors for molecular cloning
PP2389 Bacterial 6His Tag Vector Set, plasmid vectors for molecular cloning
PP2396 Bacterial C-Myc Tag Vector Set, plasmid vectors for molecular cloning
PP2385 Bacterial C-terminal Tag Vector Set, plasmid vectors for molecular cloning
PP2394 Bacterial FLAG® Tag Vector Set, plasmid vectors for molecular cloning
PP2400 Bacterial GST Tag Vector Set, plasmid vectors for molecular cloning
PP2398 Bacterial HA Tag Vector Set, plasmid vectors for molecular cloning
PP2408 Bacterial MBP Tag Vector Set, plasmid vectors for molecular cloning
PP2384 Bacterial N-terminal Tag Vector Set, plasmid vectors for molecular cloning
PP2406 Bacterial S-Tag Vector Set, plasmid vectors for molecular cloning
PP2377 Bacterial Selection Vector Set, plasmid vectors for molecular cloning
PP2380 Bacterial Signal Peptide Vector Set, plasmid vectors for molecular cloning
PP2402 Bacterial Strep Tag Vector Set, plasmid vectors for molecular cloning
PP2404 Bacterial T7 Tag Vector Set, plasmid vectors for molecular cloning
PP2392 Bacterial V5 Tag Vector Set, plasmid vectors for molecular cloning
PP2375 Blasticidin Selection Vector Set, plasmid vectors for molecular cloning
PP2370 CFP Vector Set, plasmid vectors for molecular cloning
PP2412 Cleavage Tag Vector Set, plasmid vectors for molecular cloning
PP2374 G418 Selection Vector Set, plasmid vectors for molecular cloning
PP2409 Human IgG Vector Set, plasmid vectors for molecular cloning
PP2373 Hygromycin Selection Vector Set, plasmid vectors for molecular cloning
PP2387 Mammalian 10His Tag Vector Set, plasmid vectors for molecular cloning
PP2390 Mammalian 6His Tag Vector Set, plasmid vectors for molecular cloning
PP2395 Mammalian C-Myc Tag Vector Set, plasmid vectors for molecular cloning
PP2383 Mammalian C-terminal Tag Vector Set, plasmid vectors for molecular cloning
PP2393 Mammalian FLAG® Tag Vector Set, plasmid vectors for molecular cloning
PP2399 Mammalian GST Tag Vector Set, plasmid vectors for molecular cloning
PP2397 Mammalian HA Tag Vector Set, plasmid vectors for molecular cloning
PP2407 Mammalian MBP Tag Vector Set, plasmid vectors for molecular cloning
PP2382 Mammalian N-terminal Tag Vector Set, plasmid vectors for molecular cloning