EMAIL THIS PAGE TO A FRIEND
European review for medical and pharmacological sciences
Losartan improves the distribution and efficacy of doxorubicin in CT26 tumor.
PMID 26502868
Abstract
The effectiveness of chemotherapeutic agents is impaired by limited delivery of chemotherapeutic agents to the tumor cells. Improving drug penetration in tumor tissues is very important. We tested whether losartan, a selective antagonist against type 1 angiotensin II receptors (AT1R) with noted antifibrotic activity, can enhance the penetration and efficacy of doxorubicin. BALB/C mice, which implanted with CT26 tumor cells, were divided into four groups: control, doxorubicin alone, losartan alone and doxorubicin + losartan combination groups. At day 0, the losartan alone and doxorubicin + losartan combination groups received losartan; and at day 8, the doxorubicin alone and doxorubicin + losartan combination groups received doxorubicin i.v. Tumor growth and intratumoral distribution of doxorubicin were evaluated. The mechanism underlying the enhanced anti-tumor effect of the combination of doxorubicin and losartan was investigated by immunohistochemical analysis. Treatment with losartan alone did not suppress tumor growth; In contrast, treatment with doxorubicin alone decreased tumor growth; losartan and doxorubicin were administered in combination, had a synergistic effect that the tumor growth was much more inhibited. The decreased proliferation as indicated by down-regulation of Ki67, and increased apoptosis as indicated by TUNEL and caspase-3 staining. The expression of tumor suppressor gene P53 increased in doxorubicin + losartan combination groups. Losartan can increase the therapeutic effectiveness of doxorubicin, yielding more great antitumor benefit. This study provided a rationale for initiating clinical trials using losartan in combination with chemotherapeutic agents to increase their therapeutic effectiveness.