EMAIL THIS PAGE TO A FRIEND
Nature communications
Numb is required to prevent p53-dependent senescence following skeletal muscle injury.
PMID 26503169
Abstract
Regeneration relies on coordinated action of multiple cell types to reconstitute the damaged tissue. Here we inactivate the endocytic adaptor protein Numb in skeletal muscle stem cells prior to chronic or severe muscle injury in mice. We observe two types of senescence in regenerating muscle; a transient senescence in non-myogenic cells of control and Numb mutant mice that partly depends on INK4a/ARF activity, and a persistent senescence in myogenic cells lacking Numb. The senescence levels of Numb-deficient muscle is reduced to wild type levels by an anti-oxidant treatment or p53 ablation, resulting in functional rescue of the regenerative potential in Numb mutants. Ex vivo experiments suggest that Numb-deficient senescent cells recruit macrophages to sustain inflammation and drive fibrosis, two hallmarks of the impaired muscle regeneration in Numb mutants. These findings provide insights into previously reported developmental and oncogenic senescence that are also differentially regulated by p53.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6278
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
C26H29NO2
SAB1401044
Anti-CALCR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401045
Anti-CALCR antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA028962
Anti-CALCR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700788
Anti-EMR1 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1405756
Anti-EMR1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA052809 Anti-EMR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500764
Anti-EMR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA000451
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001164
Anti-MKI67 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306667
ANTI-P53 (S15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306713
ANTI-P53 (S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306623
ANTI-P53 (SUMOYLATION SITE SPECIFIC) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1306675
ANTI-P53 (T55) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503015
Anti-p53 antibody produced in rabbit, affinity isolated antibody
P4235 Anti-p53 antibody produced in sheep, fractionated antiserum, buffered aqueous solution
SAB1306006
ANTI-P53(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503001
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503003
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503010
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503011
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503012
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503013
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503019
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503021
Anti-p53, C-Terminal antibody produced in rabbit, affinity isolated antibody
AV32393
Anti-PAX7 (AB1) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV32742
Anti-PAX7 (AB2) antibody produced in rabbit, affinity isolated antibody
SAB1412356
ANTI-PAX7 antibody produced in mouse, clone 1G11, purified immunoglobulin, buffered aqueous solution
SAB1412376
ANTI-PAX7 antibody produced in mouse, clone 3H1, purified immunoglobulin, buffered aqueous solution
SAB1412455
ANTI-PAX7 antibody produced in mouse, clone 4F8, purified immunoglobulin, buffered aqueous solution