EMAIL THIS PAGE TO A FRIEND
Oncology reports
CMTM8 inhibits the carcinogenesis and progression of bladder cancer.
PMID 26503336
Abstract
Bladder cancer is the most common tumor of the urinary tract. The incidence of bladder cancer has increased in the last few decades, thus novel molecular markers for early diagnosis and more efficacious treatment are urgently needed. Chemokine‑like factor (CKLF)‑like MARVEL transmembrane domain containing 8 (CMTM8) is downregulated in several types of cancers and is associated with tumor progression. However, CMTM8 expression has been unexplored in bladder cancer to date. Our results revealed that the expression of CMTM8 was negative in 46 of 74 (62.2%) bladder cancer samples via immunohistochemistry assay. CMTM8 downregulation was associated with advancing tumor stage and tumor grade. CMTM8 was successfully overexpressed by lentivirus in EJ and T24 cells, and the CCK‑8 and Transwell assays showed that CMTM8 overexpression decreased cell proliferation, migration and invasion in vitro. In tumor xenografts upregulation of CMTM8 inhibited tumor growth and lymph node metastasis in vivo. In conclusion, overexpression of CMTM8 in bladder cancer results in reduced malignant cell growth, migration and invasion, which could make it a potential therapeutic target in the treatment of bladder cancer.