EMAIL THIS PAGE TO A FRIEND
Anticancer research
The FGFR Inhibitor NVP-BGJ398 Induces NSCLC Cell Death by Activating Caspase-dependent Pathways as well as Caspase-independent Apoptosis.
PMID 26504010
Abstract
Fibroblast growth factor receptors are expressed in diverse cell types. They play a critical role in tumor development. Their activation promotes cell-cycle progression, angiogenesis, and cell survival by induction/suppression of the expression of proteins involved. Non-small cell lung cancer (NSCLC) cells (line H1581) were treated with NVP-BGJ398 to evaluate effects on growth by western blot, 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazoliumbromide assay and cell-cycle analysis. NVP-BGJ398 induced cell death in H1581 cells by activating caspase-dependent mitochondrial and non-mitochondrial pathways. Caspase-independent apoptosis was also activated. Cells were found to be arrested in the G0/G1 phase. Furthermore, the expression of the tumor-suppressor gene programmed cell death 4 (PDCD4) was up-regulated with suppression of angiopoietin 2 (ANG2). This represents an additional mechanism by which NVP-BGJ389 inhibits tumor growth. Various pathways induce apoptosis in NSCLC cells by employing NVP-BGJ398. These data reflect the potential of cancer treatment utilizing small FGFR inhibitors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M86804
N-Methylurea, 97%
C2H6N2O