EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Quality control of mitochondrial protein synthesis is required for membrane integrity and cell fitness.
PMID 26504172
Abstract
Mitochondrial ribosomes synthesize a subset of hydrophobic proteins required for assembly of the oxidative phosphorylation complexes. This process requires temporal and spatial coordination and regulation, so quality control of mitochondrial protein synthesis is paramount to maintain proteostasis. We show how impaired turnover of de novo mitochondrial proteins leads to aberrant protein accumulation in the mitochondrial inner membrane. This creates a stress in the inner membrane that progressively dissipates the mitochondrial membrane potential, which in turn stalls mitochondrial protein synthesis and fragments the mitochondrial network. The mitochondrial m-AAA protease subunit AFG3L2 is critical to this surveillance mechanism that we propose acts as a sensor to couple the synthesis of mitochondrial proteins with organelle fitness, thus ensuring coordinated assembly of the oxidative phosphorylation complexes from two sets of ribosomes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
40725
1,3-Dimethyl-2-imidazolidinone, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
C5H10N2O
40727
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
C5H10N2O
193453
1,3-Dimethyl-2-imidazolidinone, reagent grade
C5H10N2O
D27802
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
C6H12N2
A6671
Actinonin
C19H35N3O5
SAB4502786
Anti-MRPL14, C-Terminal antibody produced in rabbit, affinity isolated antibody
295000
Argon, ≥99.998%
Ar
601799
Argon-40Ar, 99.95 atom %
40Ar
B0126
Bromophenol Blue
C19H10Br4O5S
114391
Bromophenol Blue, ACS reagent
C19H10Br4O5S
C0378
Chloramphenicol, ≥98% (HPLC)
C11H12Cl2N2O5
C7795
Chloramphenicol, γ-irradiated
C11H12Cl2N2O5
C1919
Chloramphenicol, BioReagent, suitable for plant cell culture
C11H12Cl2N2O5
C0857
Chloramphenicol, meets USP testing specifications
C11H12Cl2N2O5
C1863
Chloramphenicol, Biotechnology Performance Certified, suitable for plant cell culture
C11H12Cl2N2O5
290734
Dabco® 33-LV
C6H12N2
861677
DL-Cysteine, technical grade
C3H7NO2S
EHU029131 MISSION® esiRNA, esiRNA human ATP5B (esiRNA1)
EMU006481 MISSION® esiRNA, esiRNA targeting mouse Atp5b (esiRNA1)
T5428
Tetramethylrhodamine methyl ester perchlorate, ≥95%
C25H25ClN2O7
V0627
Valinomycin, ≥98% (TLC), ≥90% (HPLC), solid
C54H90N6O18
94675
Valinomycin, ≥98.0% (TLC)
C54H90N6O18
V3639
Valinomycin, Ready Made Solution, ~1 mg/mL in DMSO, 0.2 μm filtered
C54H90N6O18