EMAIL THIS PAGE TO A FRIEND
Clinical drug investigation
Antiplatelet Efficacy of Fixed-Dose Aspirin-Clopidogrel Combination in Patients with Stable Coronary Artery Disease Treated with Drug-Eluting Stent Implantation.
PMID 26507618
Abstract
A fixed-dose combination (FDC) of aspirin and clopidogrel bisulfate may improve medication adherence. However, the absence of data on the relative antiplatelet efficacy of FDC and separate dual pills (SDP) of aspirin and clopidogrel in real-world patients with stable coronary artery disease is a major factor retarding clinical introduction of such an FDC. This was a single-centre, randomized, open-label, parallel-group, non-inferiority trial. Patients who maintained a regimen of separate aspirin and clopidogrel pills for at least 1xa0year after drug-eluting stent implantation without adverse events were enrolled. Patients were randomly assigned to either the FDC group or the SDP group. Antiplatelet efficacy and tolerability were assessed at baseline and at 4xa0weeks. Of the 93 enrolled patients, 83 (FDC group: nxa0=xa042; SDP group: nxa0=xa041) completed the study. The difference in the changes in P2Y12 percentage inhibition did not exceed the predetermined value for inferiority [mean difference -1.7; 95xa0% confidence interval (CI) -6.9 to 4.5, pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0004
(S)-(+)-Clopidogrel hydrogensulfate, ≥98% (HPLC)
C16H16ClNO2S · H2SO4