EMAIL THIS PAGE TO A FRIEND
Molecular biology of the cell
Angiotensin II down-regulates nephrin-Akt signaling and induces podocyte injury: roleof c-Abl.
PMID 26510503
Abstract
Recent studies have shown that nephrin plays a vital role in angiotensin II (Ang II)-induced podocyte injury and thus contributes to the onset of proteinuria and the progression of renal diseases, but its specific mechanism remains unclear. c-Abl is an SH2/SH3 domain-containing nonreceptor tyrosine kinase that is involved in cell survival and regulation of the cytoskeleton. Phosphorylated nephrin is able to interact with molecules containing SH2/SH3 domains, suggesting that c-Abl may be a downstream molecule of nephrin signaling. Here we report that Ang II-infused rats developed proteinuria and podocyte damage accompanied by nephrin dephosphorylation and minimal interaction between nephrin and c-Abl. In vitro, Ang II induced podocyte injury and nephrin and Akt dephosphorylation, which occurred in tandem with minimal interaction between nephrin and c-Abl. Moreover, Ang II promoted c-Abl phosphorylation and interaction between c-Abl and SH2 domain-containing 5'-inositol phosphatase 2 (SHIP2). c-Abl small interfering RNA (siRNA) and STI571 (c-Abl inhibitor) provided protection against Ang II-induced podocyte injury, suppressed the Ang II-induced c-Abl-SHIP2 interaction and SHIP2 phosphorylation, and maintained a stable level of nephrin phosphorylation. These results indicate that c-Abl is a molecular chaperone of nephrin signaling and the SHIP2-Akt pathway and that the released c-Abl contributes to Ang II-induced podocyte injury.
Related Materials