EMAIL THIS PAGE TO A FRIEND
Journal of industrial microbiology & biotechnology
Controlling the transcription levels of argGH redistributed L-arginine metabolic flux in N-acetylglutamate kinase and ArgR-deregulated Corynebacterium crenatum.
PMID 26521658
Abstract
Corynebacterium crenatum SYPA5-5, an L-arginine high-producer obtained through multiple mutation-screening steps, had been deregulated by the repression of ArgR that inhibits L-arginine biosynthesis at genetic level. Further study indicated that feedback inhibition of SYPA5-5 N-acetylglutamate kinase (CcNAGK) by L-arginine, as another rate-limiting step, could be deregulated by introducing point mutations. Here, we introduced two of the positive mutations (H268N or R209A) of CcNAGK into the chromosome of SYPA5-5, however, resulting in accumulation of large amounts of the intermediates (L-citrulline and L-ornithine) and decreased production of L-arginine. Genetic and enzymatic levels analysis involved in L-arginine biosynthetic pathway of recombinants SYPA5-5-NAGKH268N (H-7) and SYPA5-5-NAGKR209A (R-8) showed that the transcription levels of argGH decreased accompanied with the reduction of argininosuccinate synthase and argininosuccinase activities, respectively, which led to the metabolic obstacle from L-citrulline to L-arginine. Co-expression of argGH with exogenous plasmid in H-7 and R-8 removed this bottleneck and increased L-arginine productivity remarkably. Compared with SYPA5-5, fermentation period of H-7/pDXW-10-argGH (H-7-GH) reduced to 16 h; meanwhile, the L-arginine productivity improved about 63.6%. Fed-batch fermentation of H-7-GH in 10 L bioreactor produced 389.9 mM L-arginine with the productivity of 5.42 mM h(-1). These results indicated that controlling the transcription of argGH was a key factor for regulating the metabolic flux toward L-arginine biosynthesis after deregulating the repression of ArgR and feedback inhibition of CcNAGK, and therefore functioned as another regulatory mode for L-arginine production. Thus, deregulating all these three regulatory modes was a powerful strategy to construct L-arginine high-producing C. crenatum.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G1278
Galantide, ≥90% (HPLC)
C104H151N25O26S