EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Preparation and evaluation of amoxicillin loaded dual molecularly imprinted nanoparticles for anti-Helicobacter pylori therapy.
PMID 26524712
Abstract
This paper reports studies on preparation and evaluation of amoxicillin loaded dual molecularly imprinted nanoparticles (Amo/Dual-MIPs) designed for anti-H. pylori therapy. Both MNQA and AmoNa were chosen as templates to prepare Dual-MIPs using inverse microemulsion polymerization method. NQA was modified with myristic acid (MNQA) to become amphiphilic and assist in leaving NQA cavities on the surface of Dual-MIPs for H. pylori adhesion. AmoNa was applied to produce imprinting sites in Dual-MIPs for rebinding AmoNa to exert its anti-H. pylori effect. Batch rebinding test demonstrated a preferential rebinding effect of NQA toward the Dual-MIPs. In vivofluorescence imaging showed the prolonged residence time of Dual-MIPs in H. pylori infected mice stomachs after intragastric administration of nanoparticles.In vivo H. pylori clearance tests indicated Amo/Dual-MIPs had a better aniti-H. pylori effect than amoxicillin powder did. In conclusion, Amo/Dual-MIPs may provide an alternative drug delivery strategy for anti-H. pylori therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
543292
IR-783, Dye content 90 %
C38H46ClN2NaO6S2