EMAIL THIS PAGE TO A FRIEND
Trends in microbiology
Programming Bacteriophages by Swapping Their Specificity Determinants.
PMID 26526502
Abstract
Bacteriophages, bacteria's natural enemies, may serve as potent antibacterial agents. Their specificity for certain bacterial sub-species limits their effectiveness, but allows selective targeting of bacteria. Lu and colleagues present a platform for such targeting through alteration of bacteriophages' host specificity by swapping specificity domains in their host-recognition ligand.