EMAIL THIS PAGE TO A FRIEND
Scientific reports
Neurotoxicity induced by zinc oxide nanoparticles: age-related differences and interaction.
PMID 26527454
Abstract
This study mainly investigated the neurotoxicity induced by zinc oxide nanoparticle (ZnO NP) in different-aged mice and the interaction between age and ZnO NP exposure. Sixty adult and old male C57BL/6J mice were assigned to four groups based on a two-factor (age and ZnO NP exposure) design. Results showed that ZnO NPs (5.6 mg/kg, intraperitoneal) induced increased production of pro-inflammatory cytokines in the serum and the brain of mice. A synergistic reaction between aging and ZnO NP exposure occurred regarding serum interleukin 1 (IL-1) and interleukin 6 (IL-6). In the brain, increased oxidative stress level, impaired learning and memory abilities, and hippocampal pathological changes were identified, especially in old mice, following ZnO NP exposure. Then, a potential mechanism of cognitive impairment was examined. The contents of hippocampal cAMP response element binding protein (CREB), phosphorylated CREB, synapsin I, and cAMP were decreased in an age-dependent manner, and the most substantial decrease occurred in old mice treated with ZnO NPs. These findings demonstrated for the first time that aging and ZnO NP exposure synergistically influenced systemic inflammation, and indicated old individuals were more susceptible to ZnO NP-induced neurotoxicity. One of the mechanisms might due to the supression of cAMP/CREB signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32950
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
C13H14N2
A9501
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
C10H12N5O6P
A3262
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
C10H12N5O6P · C4H11NO3
H1758
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
HCl
435570
Hydrochloric acid, reagent grade, 37%
HCl
40233
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17863), ≥32%
HCl
40253
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17823), fuming 37%, 37-38%
HCl
84429
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
HCl
H9892
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
HCl
84434
Hydrochloric acid solution, BioXtra, ~0.1 M in H2O
HCl
H0636
Hydrochloric acid solution, protein sequencing grade, liquid
HCl
2104
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
HCl
W530574
Hydrochloric acid solution, 32 wt. % in H2O, FCC
HCl
295426
Hydrogen chloride, ReagentPlus®, ≥99%
HCl
705551
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
HCl
G4251
L-Glutathione reduced, ≥98.0%
C10H17N3O6S
G6529
L-Glutathione reduced, BioXtra, ≥98.0%
C10H17N3O6S
G6013
L-Glutathione reduced, cell culture tested, BioReagent, ≥98.0%, powder
C10H17N3O6S
40104-U Methanolic HCl, 3 M HCl in methanol (3N), for GC derivatization, pkg of 1 × 2 L
HCl