EMAIL THIS PAGE TO A FRIEND
Journal of neuroimmunology
Chrysin suppresses human CD14(+) monocyte-derived dendritic cells and ameliorates experimental autoimmune encephalomyelitis.
PMID 26531689
Abstract
Chrysin, a naturally flavonoid of plant, has various biological activities. However, the effects of chrysin on dendritic cells (DCs) and multiple sclerosis (MS) remain unknown. In this study, we demonstrate that chrysin inhibited human DC differentiation, maturation, function and the expression of the Th1 cells polarizing cytokines IFN-γ and IL-12p35 form DCs. In addition, chrysin ameliorated experimental autoimmune encephalomyelitis (EAE), an animal model of MS, by reducing CNS inflammation and demyelination. Furthermore, chrysin suppressed DCs and Th1 cells in the EAE mice. Taken together, chrysin exerts anti-inflammatory and immune suppressive effects, and suggests a possible therapeutic application of chrysin in MS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C80105
Chrysin, 97%
C15H10O4