EMAIL THIS PAGE TO A FRIEND
Circulation
Pediatric Pulmonary Hypertension: Guidelines From the American Heart Association and American Thoracic Society.
PMID 26534956
Abstract
Pulmonary hypertension is associated with diverse cardiac, pulmonary, and systemic diseases in neonates, infants, and older children and contributes to significant morbidity and mortality. However, current approaches to caring for pediatric patients with pulmonary hypertension have been limited by the lack of consensus guidelines from experts in the field. In a joint effort from the American Heart Association and American Thoracic Society, a panel of experienced clinicians and clinician-scientists was assembled to review the current literature and to make recommendations on the diagnosis, evaluation, and treatment of pediatric pulmonary hypertension. This publication presents the results of extensive literature reviews, discussions, and formal scoring of recommendations for the care of children with pulmonary hypertension.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0463
BPD, ≥98% (HPLC)
C17H10O5
N7634
Nifedipine, ≥98% (HPLC), powder
C17H18N2O6
N114
Nifedipine
C17H18N2O6