EMAIL THIS PAGE TO A FRIEND
The Analyst
Quinones as novel chemiluminescent probes for the sensitive and selective determination of biothiols in biological fluids.
PMID 26535414
Abstract
Altered plasma aminothiol concentrations are thought to be a valuable risk indicator and are interestingly utilized for routine clinical diagnosis and for the monitoring of various metabolic disorders and human diseases, and accordingly there is a need for an accurate and reliable assay capable of simultaneously determining aminothiols including glutathione (GSH), N-acetylcysteine (NAC), homocysteine (Hcys), and cysteine (Cys) in human plasma. Herein, a highly sensitive, selective, and very fast HPLC-chemiluminescence (HPLC-CL) coupled method is reported, exploiting for the first time the strong nucleophilicity and high reactivity of aminothiols toward quinones for a CL assay. The unique redox-cycling capability of quinone and/or Michael addition adducts, thioether-quinone conjugates, was utilized to establish a novel analytical method based on the reaction of adducts with dithiothreitol (DTT) to liberate reactive oxygen species (ROS), which are detected by using a luminol-CL assay. Specimen preparation involved the derivatization of aminothiols with menadione (MQ) for 5 minutes at room temperature. A unique green chemistry synthesis of thioether-quinones in HEPES buffer (pH 8.5) was introduced by using our reaction methodology without needing any hazardous organic solvent or catalyst. The aminothiol-MQ adducts were separated using solid-phase extraction followed by isocratic elution on an ODS column. Linearity was observed in the range of 2.5-500, 5-500, 10-1500, and 20-2000 nM with detection limits (S/N of 3) of 3.8, 4.2, 8, and 16 (fmol per injection) for GSH, NAC, Hcys, and Cys, respectively. The method was successfully applied for the selective determination of aminothiols in human plasma from healthy people and patients with rheumatic arthritis and diabetes mellitus. The obtained results postulated the usefulness of our method for investigating the relationship between aminothiol metabolism and related human disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43816
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
C4H10O2S2
646563
DL-Dithiothreitol solution, 1 M in H2O
C4H10O2S2
RDD002
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
C8H18N2O4S
H3375
HEPES, ≥99.5% (titration)
C8H18N2O4S
H4034
HEPES, BioPerformance Certified, ≥99.5% (titration), cell culture tested
C8H18N2O4S
PHG0001
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
RES6003H-B7
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
H7523
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
C8H18N2O4S
H6147
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
C8H18N2O4S
54457
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
C8H18N2O4S
83264
HEPES buffer solution, 1 M in H2O
C8H18N2O4S
123072
Luminol, 97%
C8H7N3O2
A8511
Luminol, ≥97% (HPLC)
C8H7N3O2
225711
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
HNO3
40230
Nitric acid, semiconductor grade PURANAL (Honeywell 17818), ≥65%
HNO3
40222
Nitric acid, semiconductor grade PURANAL (Honeywell 17952), ≥69%
HNO3
40280
Nitric acid, semiconductor grade MOS PURANAL (Honeywell 17920), ≥69%
HNO3
695025
Nitric acid, reagent grade, fuming, >90%
HNO3
695041
Nitric acid, ACS reagent, ≥90.0%
HNO3
608769
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
H14NO3
426288
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
C16H36BrN
193119
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
C16H36BrN
462144
Tetrabutylammonium bromide solution, 50 wt. % in H2O
C16H36BrN