PloS one
Composition of Carotenoids and Flavonoids in Narcissus Cultivars and their Relationship with Flower Color.
PMID 26536625
Abstract
Narcissus is widely used for cut flowers and potted plants, and is one of the most important commercial bulbous flowers in the floricultural industry. In this study, ten carotenoid and eighteen flavonoid compounds from the perianths and coronas of fifteen narcissus cultivars were measured by HPLC-APCI-MS/MS and UPLC-Q-TOF-MS/MS. Among these, six carotenoids, a total of seventeen flavonols and chlorogenic acid were identified in narcissus for the first time. A multivariate analysis was used to explore the relationship between flower color and pigment composition. We found that all-trans-violaxanthin and total carotenoid content were the main factors that affected flower color. These investigations could provide a global view of flower color formation and a theoretical basis for hybridization breeding in narcissus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L010000-HH
Acetonitrile, for DNA synthesis
C2H3N
01-0650
Acetonitrile, ≥99.8%, for HPLC
C2H3N
01-0623
Acetonitrile, for residue analysis, suitable for 5000 per JIS
C2H3N
01-0645
Acetonitrile, for chromatography
C2H3N
L110001
Acetonitrile
C2H3N
L011800
Acetonitrile
C2H3N
679186
Acetonitrile solution, contains 5 % (v/v) water, 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, for HPLC
C2H3N
703664
Acetonitrile solution, NMR reference standard, 0.23 wt. % in D2O (99.9 atom % D), water 0.05 wt. %, NMR tube size 6.5 mm × 8 in.
C2H3N
H1758
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
HCl
435570
Hydrochloric acid, reagent grade, 37%
HCl
40233
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17863), ≥32%
HCl
40253
Hydrochloric acid, semiconductor grade PURANAL (Honeywell 17823), fuming 37%, 37-38%
HCl
84429
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
HCl
H9892
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
HCl
84434
Hydrochloric acid solution, BioXtra, ~0.1 M in H2O
HCl
H0636
Hydrochloric acid solution, protein sequencing grade, liquid
HCl
2104
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
HCl
W530574
Hydrochloric acid solution, 32 wt. % in H2O, FCC
HCl
295426
Hydrogen chloride, ReagentPlus®, ≥99%
HCl
705551
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
HCl
40104-U Methanolic HCl, 3 M HCl in methanol (3N), for GC derivatization, pkg of 1 × 2 L
HCl
Q4951
Quercetin, ≥95% (HPLC), solid
C15H10O7
306975
tert-Butyl methyl ether, anhydrous, 99.8%
C5H12O
675407
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
C5H12O
320196
tert-Butyl methyl ether, reagent grade, 98%
C5H12O
20256
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
C5H12O
293210
tert-Butyl methyl ether, HPLC grade, for HPLC, 99.8%
C5H12O