EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Recent Progress in Obtaining Semiconducting Single-Walled Carbon Nanotubes for Transistor Applications.
PMID 26540144
Abstract
High purity semiconducting single-walled carbon nanotubes (s-SWCNTs) with a narrow diameter distribution are required for high-performance transistors. Achieving this goal is extremely challenging because the as-grown material contains mixtures of s-SWCNTs and metallic- (m-) SWCNTs with wide diameter distributions, typically inadequate for integrated circuits. Since 2000, numerous ex situ methods have been proposed to improve the purity of the s-SWCNTs. The majority of these techniques fail to maintain the quality and integrity of the s-SWCNTs with a few notable exceptions. Here, the progress in realizing high purity s-SWCNTs in as-grown and post-processed materials is highlighted. A comparison of transistor parameters (such as on/off ratio and field-effect mobility) obtained from test structures establishes the effectiveness of various methods and suggests opportunities for future improvements.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
257850
2-(4-Biphenylyl)-5-phenyl-1,3,4-oxadiazole, 98%
C20H14N2O