EMAIL THIS PAGE TO A FRIEND
Oncotarget
Bone marrow stroma-induced resistance of chronic lymphocytic leukemia cells to arsenic trioxide involves Mcl-1 upregulation and is overcome by inhibiting the PI3Kδ or PKCβ signaling pathways.
PMID 26540567
Abstract
CLL remains an incurable disease in spite of the many new compounds being studied. Arsenic trioxide (ATO) induces apoptosis in all CLL cell types and could constitute an efficient therapy. To further explore this, we have studied the influence of stromal cells, key components of the CLL microenvironment, on the response of CLL cells to ATO. Bone marrow stromal cells induced CLL cell resistance to 2 μM ATO and led to activation of Lyn, ERK, PI3K and PKC, as well as NF-κB and STAT3. Mcl-1, Bcl-xL, and Bfl-1 were also upregulated after the co-culture. Inhibition experiments indicated that PI3K and PKC were involved in the resistance to ATO induced by stroma. Moreover, idelalisib and sotrastaurin, specific inhibitors for PI3Kδ and PKCβ, respectively, inhibited Akt phosphorylation, NF-κB/STAT3 activation and Mcl-1 upregulation, and rendered cells sensitive to ATO. Mcl-1 was central to the mechanism of resistance to ATO, since: 1) Mcl-1 levels correlated with the CLL cell response to ATO, and 2) blocking Mcl-1 expression or function with specific siRNAs or inhibitors overcame the protecting effect of stroma. We have therefore identified the mechanism involved in the CLL cell resistance to ATO induced by bone marrow stroma and show that idelalisib or sotrastaurin block this mechanism and restore sensibility to ATO. Combination of ATO with these inhibitors may thus constitute an efficient treatment for CLL.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2911
GF 109203X, synthetic, ≥90% (HPLC)
C25H24N4O2
P0042
PP2, ≥98% (HPLC)
C15H16ClN5
T3830
Triciribine hydrate, ≥97% (HPLC)
C13H16N6O4 · xH2O