EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Enzymatic Glycosylation of Phenolic Antioxidants: Phosphorylase-Mediated Synthesis and Characterization.
PMID 26540621
Abstract
Although numerous biologically active molecules exist as glycosides in nature, information on the activity, stability, and solubility of glycosylated antioxidants is rather limited to date. In this work, a wide variety of antioxidants were glycosylated using different phosphorylase enzymes. The resulting antioxidant library, containing α/β-glucosides, different regioisomers, cellobiosides, and cellotriosides, was then characterized. Glycosylation was found to significantly increase the solubility and stability of all evaluated compounds. Despite decreased radical-scavenging abilities, most glycosides were identified to be potent antioxidants, outperforming the commonly used 2,6-bis(1,1-dimethylethyl)-4-methylphenol (BHT). Moreover, the point of attachment, the anomeric configuration, and the glycosidic chain length were found to influence the properties of these phenolic glycosides.