EMAIL THIS PAGE TO A FRIEND
Arthritis research & therapy
Excessive reactive oxygen species are therapeutic targets for intervertebral disc degeneration.
PMID 26542776
Abstract
Oxidative stress has been reported to be involved in numerous human diseases, including musculoskeletal disorders such as osteoarthritis. However, the interaction between intervertebral disc (IVD) degeneration and oxidative stress is not well understood. The purpose of the present study was to elucidate the contribution of oxidative stress to IVD degeneration and the efficacy of antioxidant treatment for degenerative discs. The expression level of an oxidative stress marker, nitrotyrosine, was assessed by immunohistochemistry and Western blotting. For evaluating intracellular reactive oxygen species (ROS) levels and oxidative stress in rat annulus fibrosus (AF) cells, flow cytometry and luciferase assay with an OKD48 construct were performed. The grade of IVD degeneration was assessed by magnetic resonance imaging and histological analysis. A high frequency of nitrotyrosine-positive cells was observed in rat and human degenerative discs. mRNA expression of catabolic factors such as tumor necrosis factor-alpha (TNF-alpha), matrix metalloprotease-3 (MMP-3), and cyclooxygenase-2 (COX-2) was significantly induced by treatment with H2O2 or buthionine sulfoximine, whereas that of aggrecan, an important chondrogenic proteoglycan, was reduced in a dose-dependent manner. Treatment with mitogen-activated protein kinase (MAPK) inhibitors blocked the inductive effect of excessive ROS on COX-2 mRNA expression. Western blotting confirmed the phosphorylation of MAPKs in H2O2 and BSO-treated AF cells. Conversely, we showed that TNF-α induced oxidative stress with increased intracellular ROS levels in AF cells. Treatment with the antioxidant N-acetyl cysteine (NAC) abrogated the catabolic effect of excessive ROS and TNF-alpha in vitro. Finally, we showed that oral administration of NAC prevented IVD degeneration in rat degenerative model. A positive feedback loop was formed between excessive ROS and TNF-alpha in AF cells. Thus, oxidative stress contributes to the progression of IVD degeneration and NAC can be a therapeutic option for IVD degeneration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6250
2-Mercaptoethanol, ≥99.0%
C2H6OS
M3148
2-Mercaptoethanol, for molecular biology, for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
C2H6OS
M7154
2-Mercaptoethanol, for electrophoresis
C2H6OS
63689
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
C2H6OS
M7556 Anti-MAP Kinase 2 (ERK-2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution, ~1 mg/mL
SAB4300499
Anti-MAPK14 (Ab-182) antibody produced in rabbit, affinity isolated antibody
SAB1405665
Anti-MAPK14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV06004
Anti-MAPK14 antibody produced in rabbit, affinity isolated antibody
M0800
Anti-p38 MAP Kinase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4500491
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500492
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500493
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500490
Anti-p38 MAPK, C-Terminal antibody produced in rabbit, affinity isolated antibody
GW21175B Anti-RK antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4502906
Anti-Synaptophysin antibody produced in rabbit, affinity isolated antibody
SAB4200544
Anti-Synaptophysin antibody, Mouse monoclonal, clone SVP-38, purified from hybridoma cell culture
HPA002858
Anti-SYP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102352
Anti-SYP antibody produced in rabbit, affinity isolated antibody
SAB4502982
Anti-TNFA antibody produced in rabbit, affinity isolated antibody
T0813 Anti-Tumor Necrosis Factor-α antibody produced in goat, affinity isolated antibody, lyophilized powder
T1816 Anti-Tumor Necrosis Factor-α antibody produced in goat, affinity isolated antibody, lyophilized powder
T0938 Anti-Tumor Necrosis Factor-α antibody produced in goat, affinity isolated antibody, lyophilized powder
T8300
Anti-Tumor Necrosis Factor-α antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
316989
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
H2O2
H1009
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
H2O2
40287
Hydrogen peroxide solution, semiconductor grade MOS PURANAL (Honeywell 17937), ≥30%
H2O2
18304
Hydrogen peroxide solution, meets analytical specification of Ph. Nord., 34.5-36.5%
H2O2
M7431 Monoclonal Anti-MAP Kinase 2 (ERK-2) antibody produced in mouse, clone 1B3B9, purified immunoglobulin, buffered aqueous solution
SAB1403701
Monoclonal Anti-MAPK14, (C-terminal) antibody produced in mouse, clone 3D5, ascites fluid
S5768
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid