EMAIL THIS PAGE TO A FRIEND
Cancer genomics & proteomics
Identification of Novel Biomarker and Therapeutic Target Candidates for Diagnosis and Treatment of Follicular Adenoma.
PMID 26543076
Abstract
Follicular adenoma is a type of benign and encapsulated nodule in the thyroid gland, but some adenomas have the potential to progress to follicular carcinoma. Therefore, it is important to monitor the state and progress of follicular adenoma in the clinic and discover drug development targets for the treatment of follicular adenoma to prevent its worsening to follicular carcinoma. Currently, the study of biomarkers and therapeutic targets lacks applications of up-to-date technologies, including proteomics and bioinformatics. To discover novel protein biomarker and therapeutic target candidates, a liquid chromatography-tandem mass spectrometry approach was applied to directly compare follicular adenoma with normal thyroid tissue samples. The proteomics analysis revealed 114 protein biomarker candidates out of 1,780 identified and quantified proteins. A comprehensive approach to prioritize the biomarker candidates by category and rank revealed CD63, DDB1, TYMP, VDAC2, and DCXR as the top five biomarker candidates. Upstream regulator analysis using the Ingenuity Pathway Analysis (IPA) software discovered four therapeutic target candidates for follicular adenoma, including TGFB1, MYC, ANGPT2, and NFE2L2. This study provided biomarker and therapeutic target candidates for a follow-up study, which will facilitate monitoring and treatment of follicular adenoma.
Related Materials