EMAIL THIS PAGE TO A FRIEND
Cancer science
Long non-coding RNA urothelial cancer-associated 1 promotes bladder cancer cell migration and invasion by way of the hsa-miR-145-ZEB1/2-FSCN1 pathway.
PMID 26544536
Abstract
Numerous studies suggest that several long non-coding RNAs (lncRNAs) play critical roles in bladder cancer development and progression. Long non-coding RNA urothelial cancer-associated 1 (lncRNA-UCA1) is highly expressed in bladder cancer tissues and cells, and it has been shown to play an important role in regulating aggressive phenotypes of bladder cancer cells. However, little is known about the molecular mechanism of lncRNA-UCA1-mediated bladder cancer cell migration and invasion. Here, we show that overexpression of lncRNA-UCA1 could induce EMT and increase the migratory and invasive abilities of bladder cancer cells. Mechanistically, lncRNA-UCA1 induced EMT of bladder cancer cells by upregulating the expression levels of zinc finger E-box binding homeobox 1 and 2 (ZEB1 and ZEB2), and regulated bladder cancer cell migration and invasion by tumor suppressive hsa-miR-145 and its target gene the actin-binding protein fascin homologue 1 (FSCN1). Furthermore, we also observed a positive correlation between lncRNA-UCA1 and ZEB1/2 expression, and a negative correlation between lncRNA-UCA1 and hsa-miR-145 expression in bladder cancer specimens. Importantly, we found that lncRNA-UCA1 repressed hsa-miR-145 expression to upregulate ZEB1/2, whereas the suppression of hsa-miR-145 could upregulate lncRNA-UCA1 expression in bladder cancer cells. Moreover, the binding site for hsa-miR-145 within exons 2 and 3 of lncRNA-UCA1 contributed to the reciprocal negative regulation of lncRNA-UCA1 and hsa-miR-145. Taken together, our results identified that lncRNA-UCA1 enhances bladder cancer cell migration and invasion in part through the hsa-miR-145/ZEB1/2/FSCN1 pathway. Therefore, lncRNA-UCA1 might act as a promising therapeutic target for the invasion and metastasis of bladder cancer.