EMAIL THIS PAGE TO A FRIEND
Retrovirology
Identification and characterization of a new type of inhibitor against the human immunodeficiency virus type-1 nucleocapsid protein.
PMID 26545586
Abstract
The human immunodeficiency virus type-1 (HIV-1) nucleocapsid protein (NC) is an essential and multifunctional protein involved in multiple stages of the viral life cycle such as reverse transcription, integration of proviral DNA, and especially genome RNA packaging. For this reason, it has been considered as an attractive target for the development of new anti-HIV drugs. Although a number of inhibitors of NC have been reported thus far, the search for NC-specific and functional inhibitor(s) with a good antiviral activity continues. In this study, we report the identification of A1752, a small molecule with inhibitory action against HIV-1 NC, which shows a strong antiviral efficacy and an IC50 around 1xa0μM. A1752 binds directly to HIV-1 NC, thereby inhibiting specific chaperone functions of NC including Psi RNA dimerization and complementary trans-activation response element (cTAR) DNA destabilization, and it also disrupts the proper Gag processing. Further analysis of the mechanisms of action of A1752 also showed that it generates noninfectious viral particles with defects in uncoating and reverse transcription in the infected cells. These results demonstrate that A1752 is a specific and functional inhibitor of NC with a novel mode of action and good antiviral efficacy. Thus, this agent provides a new type of anti-HIV NC inhibitor candidate for further drug development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21144 Anti-c-Fos antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
F7799
Anti-c-Fos antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
F137 Anti-c-Fos antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
HPA018531
Anti-C-FOS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306144
ANTI-C-FOS(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA004252
Anti-CD4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004472
Anti-CD4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503583
Anti-CD4, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2501357
Anti-DCTN3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1400426
Anti-DCTN3 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA044905
Anti-DCTN3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107711
Anti-DCTN3 antibody produced in rabbit, affinity isolated antibody
SAB2500363
Anti-ERG antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1408983
Anti-ERG antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV100625
Anti-ERG antibody produced in rabbit, IgG fraction of antiserum
SAB4300699
Anti-ERG antibody produced in rabbit, affinity isolated antibody
SAB2107313
Anti-ERG antibody produced in rabbit, affinity isolated antibody
SAB2103294
Anti-ERG, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2100832
Anti-FOS (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100833
Anti-FOS (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2502116
Anti-FOS antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV100639
Anti-FOS antibody produced in rabbit, IgG fraction of antiserum
SAB2104185
Anti-FOS antibody produced in rabbit, affinity isolated antibody
SAB4500995
Anti-FOS antibody produced in rabbit, affinity isolated antibody
SAB4500993
Anti-FOS, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500996
Anti-FOS, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA005723
Anti-FSCN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700218
Anti-FSCN1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2106726
Anti-FSCN1 antibody produced in rabbit, affinity isolated antibody
SAB1406141
Anti-MPP1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution