EMAIL THIS PAGE TO A FRIEND
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
A novel N-acetyl-glucosamine lectin of Lonchocarpus araripensis attenuates acute cellular inflammation in mice.
PMID 26546610
Abstract
This study had investigated the anti-inflammatory activity of a seed lectin (LAL) isolated from Lonchocarpus araripensis. LAL was purified by affinity chromatography (chitin column) and ion exchange chromatography (DEAE-Sephacel). In vitro LAL was tested for hemagglutinating activity against rabbit erythrocytes. In vivo LAL was assessed for the anti-inflammatory activity via intravenous injection (i.v.) in Swiss mice (25-30 g; n = 6/group) in models of paw edema and peritonitis. ANOVA (p < 0.05). LAL revealed two bands of 30 and 60 kDa (SDS-PAGE) and exhibited hemagglutinating activity. LAL (10 mg/kg) inhibited the paw edema (77%) and vascular permeability (26%) induced by carrageenan, and the paw edema induced by serotonin (80%), bradykinin (49%), sodium nitroprusside (83%), TNF-α (75%) and PGE2 (64%). LAL also inhibited the neutrophil migration induced by fMLP (70%) or carrageenan (69%). The intravital microscopy showed that LAL inhibited rolling (83%) and adhesion (70%) of leukocytes. LAL anti-inflammatory effect was reversed by its association with N-acetyl-glucosamine. The nine-daily treatment with LAL (10 mg/kg; i.v.) showed no toxicity. The novel N-acetyl-D-glucosamine-binding lectin isolated from L. araripensis seeds presents anti-inflammatory effect involving the lectin domain and the inhibition of 5-HT, BK, PGE2, NO, TNF-α and leukocyte rolling and adhesion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T48402
2,2,2-Tribromoethanol, 97%
C2H3Br3O