EMAIL THIS PAGE TO A FRIEND
Archiv der Pharmazie
First Occurrence of a Furano-glycyrrhetinoate and Its Cytotoxicity.
PMID 26548762
Abstract
(18α)-Glycyrrhetinic acid (4) was prepared from (18β)-glycyrrhetinic acid (1), and the cytotoxicity of some derivatives was investigated by photometric SRB assays employing several human tumor cell lines. In summary, (18β)-1 is slightly more cytotoxic than its (18α) epimer 4, but its cytotoxicity is negligible. Higher cytotoxicity was observed for the esters 2 and 5 and for the 3-O-acetylated esters 3 and 6. Cytotoxicity was improved dramatically when the hydroxyl group at position C-3 was replaced by an amino moiety. SeO2 oxidations gave access to a novel furano-glycyrrhetinoate 15. Interestingly, its seleno analog 16 is approximately five to six times less cytotoxic for the tumor cell lines tested, and tumor/non-tumor selectivity is lost upon replacement of the oxygen by a selenium substituent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G10105
18β-Glycyrrhetinic acid, 97%
C30H46O4