EMAIL THIS PAGE TO A FRIEND
Scientific reports
Mitochondria-related miR-141-3p contributes to mitochondrial dysfunction in HFD-induced obesity by inhibiting PTEN.
PMID 26548909
Abstract
Mitochondria-related microRNAs (miRNAs) have recently emerged as key regulators of cell metabolism and can modulate mitochondrial fusion and division. In order to investigate the roles of mitochondria-related miRNAs played in obesity, we conducted comprehensive molecular analysis in vitro and in vivo. Based on high-fat-diet (HFD) induced obese mice, we found that hepatic mitochondrial function was markedly altered. Subsequently, we evaluated the expression levels of selected mitochondria-related miRNAs and found that miR-141-3p was up-regulated strikingly in HFD mice. To further verify the role of miR-141-3p in obesity, we carried out gain-and-loss-of-function study in human HepG2 cells. We found that miR-141-3p could modulate ATP production and induce oxidative stress. Through luciferase report gene assay, we identified that phosphatase and tensin homolog (PTEN) was a target of miR-141-3p. Inhibiting PTEN could alter the mitochondrial function, too. Our study suggested that mitochondria-related miR-141-3p induced mitochondrial dysfunction by inhibiting PTEN.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300337
Anti-PTEN (Ab-370) antibody produced in rabbit, affinity isolated antibody
SAB4300336
Anti-PTEN (Ab-380/382/383) antibody produced in rabbit, affinity isolated antibody
SAB2501614 Anti-PTEN antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406331
Anti-PTEN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
P7482
Anti-PTEN antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500337
Anti-PTEN antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4502473
Anti-PTEN, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502474
Anti-PTEN, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502475
Anti-PTEN, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502476
Anti-PTEN, C-Terminal antibody produced in rabbit, affinity isolated antibody
WH0005728M1
Monoclonal Anti-PTEN antibody produced in mouse, clone 2G9, purified immunoglobulin, buffered aqueous solution
P3487
Monoclonal Anti-PTEN antibody produced in mouse, ~1 mg/mL, purified immunoglobulin, clone PTN-18, buffered aqueous solution
SAB1404272
Monoclonal Anti-PTEN antibody produced in mouse, clone 3E7, purified immunoglobulin, buffered aqueous solution
SAB4100580
Monoclonal Anti-PTEN antibody produced in mouse, clone PTEN-A552, culture supernatant
PLA0133
Rabbit anti-PTEN Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.