EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Expanded metabolomics approach to profiling endogenous carbohydrates in the serum of ovarian cancer patients.
PMID 26549419
Abstract
We applied hydrophilic interaction liquid chromatography coupled with tandem mass spectrometry to the quantitative analysis of serum from 58 women, including ovarian cancer patients, ovarian benign tumor patients, and healthy controls. All of these ovarian cancer and ovarian benign tumor patients have elevated cancer antigen 125, which makes them clinically difficult to differentiate the malignant from the benign. All of the 16 endogenous carbohydrates were quantitatively detected in the human sera, of which, eight endogenous carbohydrates were significantly different (P-value < 0.05) between the ovarian cancer and healthy control. According to the receiver operating characteristic curve analysis, arabitol was the most potentially specific biomarker for discriminating ovarian cancer from healthy control, having an area under the curve of 0.911. A panel of metabolite markers composed of maltose, maltotriose, raffinose, and mannitol was selected, which was able to discriminate the ovarian cancer from the benign ovarian tumor counterparts, with an area under concentration-time curve value of 0.832. Endogenous carbohydrates in the expanded metabolomics approach after the global metabolic profiling are characterized and are potential biomarkers for the early diagnosis of ovarian cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5500
Melibiose, ≥98% (HPLC)
C12H22O11