EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Highly Elastic and Conductive Human-Based Protein Hybrid Hydrogels.
PMID 26551969
Abstract
A highly elastic hybrid hydrogel of methacryloyl-substituted recombinant human tropoelastin (MeTro) and graphene oxide (GO) nanoparticles are developed. The synergistic effect of these two materials significantly enhances both ultimate strain (250%), reversible rotation (9700°), and the fracture energy (38.8 ± 0.8 J m(-2) ) in the hybrid network. Furthermore, improved electrical signal propagation and subsequent contraction of the muscles connected by hybrid hydrogels are observed in ex vivo tests.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0706 Tropoelastin human, recombinant, expressed in E. coli, 1 mg