EMAIL THIS PAGE TO A FRIEND
Journal of food protection
Retention of Rotavirus Infectivity in Mussels Heated by Using the French Recipe Moules Marinières.
PMID 26555530
Abstract
To evaluate the persistence of infectious virus after heating, mussels contaminated with a rotavirus strain were prepared following the French recipe moules marinières (mariner's mussels). Rotavirus was then quantified by real-time quantitative PCR (RT-qPCR) and a cell culture infectivity assay. Results showed the persistence of infectious virus after 3 min of cooking. After 5 min, when no infectious virus could be detected, the RT-qPCR approach showed a 1-log decrease compared with concentrations detected after 1 min of cooking.