PloS one
Human CD180 Transmits Signals via the PIM-1L Kinase.
PMID 26555723
Abstract
Toll-like receptors (TLRs) are important sensors of the innate immune system that recognize conserved structural motifs and activate cells via a downstream signaling cascade. The CD180/MD1 molecular complex is an unusual member of the TLR family, since it lacks the components that are normally required for signal transduction by other TLRs. Therefore the CD180/MD 1 complex has been considered of being incapable of independently initiating cellular signals. Using chemogenetic approaches we identified specifically the membrane bound long form of PIM-1 kinase, PIM-1L as the mediator of CD180-dependent signaling. A dominant negative isoform of PIM-1L, but not of other PIM kinases, inhibited signaling elicited by cross-linking of CD180, and this effect was phenocopied by PIM inhibitors. PIM-1L was directed to the cell membrane by its N-terminal extension, where it colocalized and physically associated with CD180. Triggering CD180 also induced increased phosphorylation of the anti-apoptotic protein BAD in a PIM kinase-dependent fashion. Also in primary human B cells, which are the main cells expressing CD180 in man, cross-linking of CD180 by monoclonal antibodies stimulated cell survival and proliferation that was abrogated by specific inhibitors. By associating with PIM-1L, CD180 can thus obtain autonomous signaling capabilities, and this complex is then channeling inflammatory signals into B cell survival programs. Pharmacological inhibition of PIM-1 should therefore provide novel therapeutic options in diseases that respond to innate immune stimulation with subsequently increased B cell activity, such as lupus erythematosus or myasthenia gravis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA003740
Anti-CD180 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700836
Anti-CD180 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701032
Anti-CD180 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500289
Anti-RP105 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
D8284
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
C20H10N2Na2O4 · xH2O
B4429
BIS-TRIS, BioPerformance Certified, cell culture tested, suitable for insect cell culture, ≥98.0%
C8H19NO5
B9754
BIS-TRIS, ≥98.0% (titration)
C8H19NO5
RES1161B-A7
BIS-TRIS, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture
C8H19NO5
PHG0004
BIS-TRIS, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H19NO5
B7535
BIS-TRIS, BioXtra, ≥98.0% (titration)
C8H19NO5
14879
BIS-TRIS, BioUltra, ≥99.0% (NT)
C8H19NO5
RAB0244
Human IL-10 ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0249
Human IL-10 R β ELISA Kit, for serum, plasma, cell culture supernatant and urine
SAB4700748 Monoclonal Anti-CD180 antibody produced in mouse, clone G28-8, purified immunoglobulin, buffered aqueous solution
RAB0245
Mouse IL-10 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0246
Rat IL-10 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0247
Rat IL-10 ELISA Kit, for cell and tissue lysates