EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Photo-Fenton oxidation of 3-amino-5-methylisoxazole: a by-product from biological breakdown of some pharmaceutical compounds.
PMID 26555882
Abstract
The present study aims to assess the removal of 3-amino-5-methylisoxazole (AMI), a recalcitrant by-product resulting from the biological breakdown of some pharmaceuticals, applying a solar photo-Fenton process assisted by ferrioxalate complexes (SPFF) (Fe(3+)/H2O2/oxalic acid/UVA-Vis) and classical solar photo-Fenton process (SPF) (Fe(2+)/H2O2/UVA-Vis). The oxidation ability of SPFF was evaluated at different iron/oxalate molar ratios (1:3, 1:6, and 1:9, with [total iron] = 3.58 × 10(-2)xa0mM and [oxalic acid] = 1.07 × 10(-1), 2.14 × 10(-1) and 3.22 × 10(-1)xa0mM, respectively) and pH values (3.5-6.5), using low iron contents (2.0xa0mg Fe(3+) L(-1)). Additionally, the use of other organic ligands such as citrate and ethylenediamine-N,N'-disuccinic acid (EDDS) was tested. The oxidation power of the classical SPF was assessed at different pH values (2.8-4.0) using 2.0xa0mg Fe(2+) per liter. Furthermore, the effect of AMI concentration (2-20xa0mgxa0L(-1)), presence of inorganic ions (Cl(-), SO4(2-), NO3(-), HCO3(-), NH4(+)), and radical scavengers (sodium azide and D-mannitol) on the SPF method at pH 3.5 was also assessed. Experiments were done using a lab-scale photoreactor with a compound parabolic collector (CPC) under simulated solar radiation. A pilot-scale assay was conducted using the best operation conditions. While at near neutral pH, an iron/oxalate molar ratio of 1:9 led to the removal of 72xa0% of AMI after 90xa0min of SPFF, at pH 3.5, an iron/oxalate molar ratio of 1:3 was enough to achieve complete AMI degradation (below the detection limit) after 30xa0min of reaction. The SPF process at pH 3.5 underwent a slower AMI degradation, reaching total AMI degradation after 40xa0min of reaction. The scale up of SPF process showed a good reproducibility. Oxalic and oxamic acids were identified as the main low-molecular-weight carboxylic acids detected during the pilot-scale SPF reaction. Graphical abstract ᅟ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
232270
3-Amino-5-methylisoxazole, ≥97%
C4H6N2O