PloS one
A Novel Adeno-Associated Virus-Based Genetic Vaccine Encoding the Hepatitis C Virus NS3/4 Protein Exhibits Immunogenic Properties in Mice Superior to Those of an NS3-Protein-Based Vaccine.
PMID 26556235
Abstract
More than 170 million individuals worldwide are infected with hepatitis C virus (HCV), and up to an estimated 30% of chronically infected individuals will go on to develop progressive liver disease. Despite the recent advances in antiviral treatment of HCV infection, it remains a major public health problem. Thus, development of an effective vaccine is urgently required. In this study, we constructed novel adeno-associated virus (AAV) vectors expressing the full-length NS3 or NS3/4 protein of HCV genotype 1b. The expression of the NS3 or NS3/4 protein in HepG2 cells was confirmed by western blotting. C57BL/6 mice were intramuscularly immunised with a single injection of AAV vectors, and the resultant immune response was investigated. The AAV2/rh32.33.NS3/4 vaccine induced stronger humoral and cellular responses than did the AAV2/rh32.33.NS3 vaccine. Our results demonstrate that AAV-based vaccines exhibit considerable potential for the development of an effective anti-HCV vaccine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
860336
3,3′,5,5′-Tetramethylbenzidine, ≥99%
C16H20N2
T2885
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
C16H20N2
T5525
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
C16H20N2
87748
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
C16H20N2
74875
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
C24H21Na3O11S3
746487
Cesium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99.9%
ClCs
203025
Cesium chloride, ≥99.999% trace metals basis
ClCs
449733
Cesium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
ClCs
289329
Cesium chloride, ReagentPlus®, 99.9%
ClCs
C3139
Cesium chloride, optical grade, ≥99.5% trace metals basis
ClCs
C3011
Cesium chloride, Grade I, ≥99.0%
ClCs
562599
Cesium chloride, 99.99% trace metals basis
ClCs
562602
Cesium chloride, Cabot high-purity grade
ClCs
C3032
Cesium chloride, for molecular biology, ≥99% (silver nitrate titration)
ClCs
C4036
Cesium chloride, for molecular biology, ≥98%
ClCs
C3309
Cesium chloride, BioXtra, ≥99.5% (titration)
ClCs
C6914
Cesium chloride, Grade II, ≥98%
ClCs
20966
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
ClCs
31807
Cesium chloride, puriss. p.a., ≥99.5%
ClCs
D1435
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
C2H6OS
07-4860
Dimethyl sulfoxide, JIS special grade, ≥99.0%
C2H6OS
07-4871
Dimethyl sulfoxide, for HPLC
C2H6OS
606987
Dimethyl sulfoxide-12C2, 99.95 atom % 12C
C2H6SO
M0503
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
C15H18N4O5
M4287
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
C15H18N4O5
M0440
Mitomycin C from Streptomyces caespitosus, ≥970 μg/mg (USP XXIV)
C15H18N4O5
M7949
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), potency: ≥970 μg per mg (USP XXIV), γ-irradiated, suitable for cell culture
C15H18N4O5
M2128
Thiazolyl Blue Tetrazolium Bromide, 98%
C18H16BrN5S
M5655
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
C18H16BrN5S