EMAIL THIS PAGE TO A FRIEND
Dermatosen in Beruf und Umwelt. Occupation and environment
[Therapy of hydrofluoric acid burns].
PMID 2656173
Abstract
Burns caused by exposure to hydrofluoric acid require immediate First Aid and specific therapeutic measures by the first available physician. Based on a review of the literature the following outline of therapy is proposed.