EMAIL THIS PAGE TO A FRIEND
Scientific reports
Dynamic interplay of multidrug transporters with TolC for isoprenol tolerance in Escherichia coli.
PMID 26563610
Abstract
Engineering of efflux pumps is a promising way to improve host's tolerance to biofuels such as medium-chain alcohols (CmOHs); however, this strategy is restricted by poor understanding of the efflux pumps engaged in extrusion of solvents. In this study, several Escherichia coli mutants of multidrug transporters were evaluated for isoprenol tolerance. Susceptible phenotypes were observed in the mutants with individual deletion of six transporters, AcrD, EmrAB, MacAB, MdtBC, MdtJI and YdiM, whereas inactivation of AcrAB transporter resulted in an improved tolerance to isoprenol and other CmOHs. AcrAB is the major transporter forming tripartite transperiplasmic complex with outer membrane channel TolC for direct extrusion of toxic molecules in E. coli. The AcrAB inactivation enables to enhance TolC availability for the multidrug transporters associated with extrusion of CmOHs and increase the tolerance to CmOHs including isoprenol. It is assumed that outer membrane channel TolC plays an important role in extrusion of isoprenol and other CmOHs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B7906
1-Butanol, for molecular biology, ≥99%
C4H10O
281549
1-Butanol, anhydrous, 99.8%
C4H10O
154679
1-Butanol, spectrophotometric grade, 99.5%
C4H10O
270679
1-Butanol, for HPLC, 99.8%
C4H10O
442665
1-Butanol, analytical standard
C4H10O
03-4585
1-Butanol, for HPLC
C4H10O
471402
1-Hexanol, anhydrous, ≥99%
C6H14O
H13303
1-Hexanol, reagent grade, 98%
C6H14O
52828
1-Hexanol, ReagentPlus®, ≥99.5% (GC)
C6H14O
398268
1-Pentanol, ACS reagent, ≥99%
C5H12O
138975
1-Pentanol, ReagentPlus®, ≥99%
C5H12O
76929
1-Pentanol, puriss. p.a., ACS reagent, ≥99.0% (GC)
C5H12O
309435
3-Methyl-1-butanol, anhydrous, ≥99%
C5H12O
320021
3-Methyl-1-butanol, ACS reagent, ≥98.5%
C5H12O
M32658
3-Methyl-1-butanol, reagent grade, 98%
C5H12O
I9392
3-Methylbutanol, BioReagent, for molecular biology, ≥98.5%
C5H12O
59085
3-Methylbutanol, BioUltra, for molecular biology, ≥99.0% (GC)
C5H12O
59090
3-Methylbutanol, puriss. p.a., ACS reagent, ≥98.5% (GC)
C5H12O
W205605
Amyl alcohol, ≥99%, FG
C5H12O
W217816
Butyl alcohol, natural, ≥99.5%, FCC, FG
C4H10O
W217808
Butyl alcohol, ≥99.9%, FCC
C4H10O
W256706
Hexyl alcohol, FCC, FG
C6H14O
W256714
Hexyl alcohol, natural, ≥98%, FCC, FG
C6H14O
W205702
Isoamyl alcohol, ≥98%, FG
C5H12O
W205710
Isoamyl alcohol, natural, ≥98%, FG
C5H12O
T0397
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
C15H10I4NNaO4 · 5H2O
T2501
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
C15H10I4NNaO4 · 5H2O