EMAIL THIS PAGE TO A FRIEND
Shock (Augusta, Ga.)
Hydrogen-Rich Saline Attenuates Lipopolysaccharide-Induced Heart Dysfunction by Restoring Fatty Acid Oxidation in Rats by Mitigating C-Jun N-Terminal Kinase Activation.
PMID 26565704
Abstract
Sepsis is common in intensive care units (ICU) and is associated with high mortality. Cardiac dysfunction complicating sepsis is one of the most important causes of this mortality. This dysfunction is due to myocardial inflammation and reduced production of energy by the heart. A number of studies have shown that hydrogen-rich saline (HRS) has a beneficial effect on sepsis. Therefore, we tested whether HRS prevents cardiac dysfunction by increasing cardiac energy. Four groups of rats received intraperitoneal injections of one of the following solutions: normal saline (NS), HRS, lipopolysaccharide (LPS), and LPS plus HRS. Cardiac function was measured by echocardiography 8 h after the injections. Gene and protein expression related to fatty acid oxidation (FAO) were measured by quantitative polymerase chain reaction (PCR) and Western blot analysis. The injection of LPS compromised heart function through decreased fractional shortening (FS) and increased left ventricular diameter (LVD). The addition of HRS increased FS, palmitate triphosphate, and the ratio of phosphocreatinine (PCr) to adenosine triphosphate (ATP) as well as decreasing LVD. The LPS challenge reduced the expression of genes related to FAO, including perioxisome proliferator-activated receptor gamma coactivator 1-alpha (PGC-1α), perioxisome proliferator-activated receptor alpha (PPARα), Estrogen-related receptor alpha (ERRα), and their downstream targets, in mRNA and protein level, which were attenuated by HRS. However, HRS had little effect on glucose metabolism. Furthermore, HRS inhibited c-Jun N-terminal kinase (JNK) activation in the rat heart. Inhibition of JNK by HRS showed beneficial effects on LPS-challenged rats, at least in part, by restoring cardiac FAO.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA011990
Anti-ACADL antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV33855
Anti-ACADL antibody produced in rabbit, affinity isolated antibody
SAB2100020
Anti-ACADL antibody produced in rabbit, affinity isolated antibody
SAB2500017
Anti-ACADM antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405417
Anti-ACADM antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA006198
Anti-ACADM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32788
Anti-ACADM antibody produced in rabbit, affinity isolated antibody
AV32789
Anti-ACADM antibody produced in rabbit, IgG fraction of antiserum
HPA026542
Anti-ACADM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100714
Anti-ESRRA antibody produced in rabbit, affinity isolated antibody
SAB2106252
Anti-ESRRA antibody produced in rabbit, affinity isolated antibody
SAB1410347
Anti-ESRRA antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
E0406 Anti-Estrogen-Related Receptor α antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4200176
Anti-JNK antibody, Mouse monoclonal, clone 1C2, purified from hybridoma cell culture
SAB4502398
Anti-JNK1 antibody produced in rabbit, affinity isolated antibody
SAB1406300
Anti-MAPK8 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1101208
Anti-PGC (151-164) antibody produced in rabbit, IgG fraction of antiserum
SAB1400204
Anti-PGC antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1401284
Anti-PGC antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA031718
Anti-PGC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031717
Anti-PGC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1402105 Monoclonal Anti-ACADM, (N-terminal) antibody produced in mouse, clone 3A2, purified immunoglobulin, buffered aqueous solution
WH0005599M2
Monoclonal Anti-MAPK8 antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution
SAB1404249
Monoclonal Anti-MAPK8 antibody produced in mouse, clone 2F11, purified immunoglobulin, buffered aqueous solution