PloS one
Identification of New Antifungal Compounds Targeting Thioredoxin Reductase of Paracoccidioides Genus.
PMID 26569405
Abstract
The prevalence of invasive fungal infections worldwide has increased in the last decades. The development of specific drugs targeting pathogenic fungi without producing collateral damage to mammalian cells is a daunting pharmacological challenge. Indeed, many of the toxicities and drug interactions observed with contemporary antifungal therapies can be attributed to "nonselective" interactions with enzymes or cell membrane systems found in mammalian host cells. A computer-aided screening strategy against the TRR1 protein of Paracoccidioides lutzii is presented here. Initially, a bank of commercially available compounds from Life Chemicals provider was docked to model by virtual screening simulations. The small molecules that interact with the model were ranked and, among the best hits, twelve compounds out of 3,000 commercially-available candidates were selected. These molecules were synthesized for validation and in vitro antifungal activity assays for Paracoccidioides lutzii and P. brasiliensis were performed. From 12 molecules tested, 3 harbor inhibitory activity in antifungal assays against the two pathogenic fungi. Corroborating these findings, the molecules have inhibitory activity against the purified recombinant enzyme TRR1 in biochemical assays. Therefore, a rational combination of molecular modeling simulations and virtual screening of new drugs has provided a cost-effective solution to an early-stage medicinal challenge. These results provide a promising technique to the development of new and innovative drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74875
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
C24H21Na3O11S3
D2650
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
C2H6OS
D8418
Dimethyl sulfoxide, for molecular biology
C2H6OS
D4540
Dimethyl sulfoxide, ≥99.5% (GC), plant cell culture tested
C2H6OS
D1435
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
C2H6OS
D9170
Dimethyl sulfoxide, PCR Reagent
C2H6OS
276855
Dimethyl sulfoxide, anhydrous, ≥99.9%
C2H6OS
41639
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
C2H6OS
07-4860
Dimethyl sulfoxide, JIS special grade, ≥99.0%
C2H6OS
07-4871
Dimethyl sulfoxide, for HPLC
C2H6OS
673439 Dimethyl sulfoxide solution, 50 wt. % in H2O
C2H6OS
606987
Dimethyl sulfoxide-12C2, 99.95 atom % 12C
C2H6SO
F8929
Fluconazole, ≥98% (HPLC), powder
C13H12F2N6O
RDD003
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
C7H15NO4S
M1254
MOPS, ≥99.5% (titration)
C7H15NO4S
PHG0007
MOPS, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C7H15NO4S
M3183
MOPS, BioPerformance Certified, cell culture tested, ≥99.5% (titration)
C7H15NO4S
M5162
MOPS, BioXtra, ≥99.5% (titration)
C7H15NO4S
69948
MOPS, BioUltra, ≥99.5% (T)
C7H15NO4S