EMAIL THIS PAGE TO A FRIEND
Journal of investigative medicine : the official publication of the American Federation for Clinical Research
Lowering Uric Acid With Allopurinol Improves Insulin Resistance and Systemic Inflammation in Asymptomatic Hyperuricemia.
PMID 26571421
Abstract
Hyperuricemia is an independent predictor of impaired fasting glucose and type 2 diabetes, but whether it has a causal role in insulin resistance remains controversial. Here we tested the hypothesis that lowering uric acid in hyperuricemic nondiabetic subjects might improve insulin resistance. Subjects with asymptomatic hyperuricemia (n = 73) were prospectively placed on allopurinol (n = 40) or control (n = 33) for 3 months. An additional control group consisted of 48 normouricemic subjects. Serum uric acid, fasting glucose, fasting insulin, HOMA-IR (homeostatic model assessment of insulin resistance), and high-sensitivity C-reactive protein were measured at baseline and at 3 months. Allopurinol-treated subjects showed a reduction in serum uric acid in association with improvement in fasting blood glucose, fasting insulin, and HOMA-IR index, as well as a reduction in serum high-sensitivity C-reactive protein. The number of subjects with impaired fasting glucose significantly decreased in the allopurinol group at 3 months compared with baseline (n = 8 [20%] vs n = 30 [75%], 3 months vs baseline, P < 0.001). In the hyperuricemic control group, only glucose decreased significantly and, in the normouricemic control, no end point changed. Allopurinol lowers uric acid and improves insulin resistance and systemic inflammation in asymptomatic hyperuricemia. Larger clinical trials are recommended to determine if lowering uric acid can help prevent type 2 diabetes.