PloS one
SWI/SNF Subunits SMARCA4, SMARCD2 and DPF2 Collaborate in MLL-Rearranged Leukaemia Maintenance.
PMID 26571505
Abstract
Alterations in chromatin structure caused by deregulated epigenetic mechanisms collaborate with underlying genetic lesions to promote cancer. SMARCA4/BRG1, a core component of the SWI/SNF ATP-dependent chromatin-remodelling complex, has been implicated by its mutational spectrum as exerting a tumour-suppressor function in many solid tumours; recently however, it has been reported to sustain leukaemogenic transformation in MLL-rearranged leukaemia in mice. Here we further explore the role of SMARCA4 and the two SWI/SNF subunits SMARCD2/BAF60B and DPF2/BAF45D in leukaemia. We observed the selective requirement for these proteins for leukaemic cell expansion and self-renewal in-vitro as well as in leukaemia. Gene expression profiling in human cells of each of these three factors suggests that they have overlapping functions in leukaemia. The gene expression changes induced by loss of the three proteins demonstrate that they are required for the expression of haematopoietic stem cell associated genes but in contrast to previous results obtained in mouse cells, the three proteins are not required for the expression of c-MYC regulated genes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV38768
Anti-DPF2 antibody produced in rabbit, IgG fraction of antiserum
HPA020880
Anti-DPF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100615
Anti-DPF2 antibody produced in rabbit, affinity isolated antibody
SAB2104037
Anti-DPF2 antibody produced in rabbit, affinity isolated antibody
SAB2104355
Anti-DPF2 antibody produced in rabbit, affinity isolated antibody
AV38223
Anti-SMARCD2 antibody produced in rabbit, affinity isolated antibody
SAB2104040
Anti-SMARCD2 antibody produced in rabbit, affinity isolated antibody
SAB2104041
Anti-SMARCD2 antibody produced in rabbit, affinity isolated antibody
HPA002131
Anti-VCL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
V4139 Anti-Vinculin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503069
Anti-Vinculin antibody produced in rabbit, affinity isolated antibody
V9264
Anti-Vinculin antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture
SAB4200195
Monoclonal Anti-Brg1 antibody produced in rat, ~1.0 mg/mL, clone 5B7, purified immunoglobulin
WH0005977M1
Monoclonal Anti-DPF2 antibody produced in mouse, clone 2F6, purified immunoglobulin, buffered aqueous solution
WH0006603M1
Monoclonal Anti-SMARCD2 antibody produced in mouse, clone 2F7, purified immunoglobulin, buffered aqueous solution
SAB1404379
Monoclonal Anti-SMARCD2 antibody produced in mouse, clone 2B2, purified immunoglobulin, buffered aqueous solution
SAB1409685
Monoclonal Anti-SMARCD2 antibody produced in mouse, clone 2C2, purified immunoglobulin, buffered aqueous solution
SAB1404522
Monoclonal Anti-VCL antibody produced in mouse, clone 3F8-1D4, purified immunoglobulin, buffered aqueous solution
SAB4200080
Monoclonal Anti-Vinculin antibody produced in mouse, clone V284, purified immunoglobulin, buffered aqueous solution
V4505
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid